Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50541625-50541850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN3
Entrez Gene ID
55040
Gene Name
epsin 3
Gene Aliases
-
UniGene
Hs.670090
Gene Chromosome Location
Chr.17: 50532687-50543750 on Build GRCh38
Transcripts
NM_017957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067664 c.1645A>G p.K549E Chr.17: 50541670-50541670 on Build GRCh38
4067663 c.1561A>G p.K521E Chr.17: 50541670-50541670 on Build GRCh38
7154718 c.1575C>T p.P525P Chr.17: 50541684-50541684 on Build GRCh38
7189188 c.1607T>G p.V536G Chr.17: 50541632-50541632 on Build GRCh38
7189187 c.1523T>G p.V508G Chr.17: 50541632-50541632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763344117 Chr.17: 50541705-50541705 on Build GRCh38
rs113914602 Chr.17: 50541814-50541814 on Build GRCh38
rs749556056 Chr.17: 50541730-50541732 on Build GRCh38
rs753237368 Chr.17: 50541734-50541734 on Build GRCh38
rs914624793 Chr.17: 50541792-50541792 on Build GRCh38
rs550762930 Chr.17: 50541693-50541693 on Build GRCh38
rs956167355 Chr.17: 50541711-50541711 on Build GRCh38
rs564613594 Chr.17: 50541741-50541741 on Build GRCh38
rs756932117 Chr.17: 50541746-50541746 on Build GRCh38
rs746903649 Chr.17: 50541680-50541680 on Build GRCh38
rs745620029 Chr.17: 50541648-50541648 on Build GRCh38
rs774981006 Chr.17: 50541719-50541719 on Build GRCh38
rs533127056 Chr.17: 50541752-50541752 on Build GRCh38
rs997093106 Chr.17: 50541756-50541756 on Build GRCh38
rs577978019 Chr.17: 50541665-50541665 on Build GRCh38
rs1056349979 Chr.17: 50541665-50541665 on Build GRCh38
rs562320157 Chr.17: 50541682-50541682 on Build GRCh38
rs759929471 Chr.17: 50541724-50541724 on Build GRCh38
rs748202261 Chr.17: 50541747-50541747 on Build GRCh38
rs758577490 Chr.17: 50541793-50541793 on Build GRCh38
rs779905916 Chr.17: 50541673-50541673 on Build GRCh38
rs12937447 Chr.17: 50541668-50541668 on Build GRCh38
rs779705283 Chr.17: 50541816-50541816 on Build GRCh38
rs781357104 Chr.17: 50541829-50541829 on Build GRCh38
rs911216888 Chr.17: 50541802-50541802 on Build GRCh38
rs773745424 Chr.17: 50541699-50541699 on Build GRCh38
rs1029925809 Chr.17: 50541759-50541759 on Build GRCh38
rs778095939 Chr.17: 50541712-50541712 on Build GRCh38
rs779202234 Chr.17: 50541645-50541645 on Build GRCh38
rs758334079 Chr.17: 50541654-50541654 on Build GRCh38
rs148199218 Chr.17: 50541696-50541696 on Build GRCh38
rs763708722 Chr.17: 50541733-50541733 on Build GRCh38
rs200979323 Chr.17: 50541690-50541690 on Build GRCh38
rs139485679 Chr.17: 50541796-50541796 on Build GRCh38
rs765532804 Chr.17: 50541674-50541681 on Build GRCh38
rs754853206 Chr.17: 50541820-50541820 on Build GRCh38
rs767008249 Chr.17: 50541712-50541712 on Build GRCh38
rs896148124 Chr.17: 50541666-50541666 on Build GRCh38
rs200160943 Chr.17: 50541819-50541819 on Build GRCh38

View Full Product Details