Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15297611-15297860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB11
Entrez Gene ID
140456
Gene Name
ankyrin repeat and SOCS box containing 11
Gene Aliases
-
UniGene
Hs.352183
Gene Chromosome Location
Chr.X: 15281697-15315624 on Build GRCh38
Transcripts
NM_001012428.2 NM_001201583.1 NM_080873.2

SNP Detail

SNP ID Chromosome Location
rs756939281 Chr.X: 15297659-15297659 on Build GRCh38
rs765173933 Chr.X: 15297664-15297664 on Build GRCh38
rs756213002 Chr.X: 15297729-15297729 on Build GRCh38
rs145809121 Chr.X: 15297667-15297667 on Build GRCh38
rs141242752 Chr.X: 15297633-15297633 on Build GRCh38
rs943155862 Chr.X: 15297791-15297791 on Build GRCh38
rs760107192 Chr.X: 15297634-15297634 on Build GRCh38
rs931508478 Chr.X: 15297747-15297747 on Build GRCh38
rs1049247517 Chr.X: 15297749-15297749 on Build GRCh38
rs910490355 Chr.X: 15297756-15297756 on Build GRCh38
rs1007629677 Chr.X: 15297833-15297833 on Build GRCh38
rs887574140 Chr.X: 15297755-15297755 on Build GRCh38
rs897287900 Chr.X: 15297722-15297722 on Build GRCh38
rs187336193 Chr.X: 15297820-15297820 on Build GRCh38
rs913609870 Chr.X: 15297666-15297666 on Build GRCh38
rs373751514 Chr.X: 15297672-15297672 on Build GRCh38
rs775130448 Chr.X: 15297631-15297631 on Build GRCh38
rs746910757 Chr.X: 15297709-15297709 on Build GRCh38
rs146811336 Chr.X: 15297680-15297680 on Build GRCh38
rs753567517 Chr.X: 15297650-15297650 on Build GRCh38
rs1037305516 Chr.X: 15297711-15297712 on Build GRCh38
rs757138807 Chr.X: 15297648-15297648 on Build GRCh38
rs762369980 Chr.X: 15297736-15297736 on Build GRCh38
rs758430401 Chr.X: 15297674-15297674 on Build GRCh38

View Full Product Details