Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37964368-37964605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
UniGene
Hs.475963
Gene Chromosome Location
Chr.3: 37862152-37984469 on Build GRCh38
Transcripts
NM_001008392.1 NM_005808.2 XM_017005519.1 XM_017005520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446419 c.292G>A p.E98K Chr.3: 37964595-37964595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746812352 Chr.3: 37964525-37964525 on Build GRCh38
rs375692687 Chr.3: 37964558-37964558 on Build GRCh38
rs1032539850 Chr.3: 37964582-37964582 on Build GRCh38
rs768483176 Chr.3: 37964542-37964542 on Build GRCh38
rs1892812 Chr.3: 37964521-37964521 on Build GRCh38
rs769831114 Chr.3: 37964553-37964553 on Build GRCh38
rs577081809 Chr.3: 37964479-37964479 on Build GRCh38
rs151189162 Chr.3: 37964577-37964577 on Build GRCh38
rs377485791 Chr.3: 37964537-37964537 on Build GRCh38
rs545849444 Chr.3: 37964481-37964481 on Build GRCh38
rs1006564100 Chr.3: 37964407-37964407 on Build GRCh38
rs773003699 Chr.3: 37964556-37964556 on Build GRCh38
rs770719007 Chr.3: 37964446-37964446 on Build GRCh38
rs547909829 Chr.3: 37964404-37964404 on Build GRCh38
rs748677776 Chr.3: 37964445-37964445 on Build GRCh38
rs138952929 Chr.3: 37964472-37964472 on Build GRCh38
rs1023952189 Chr.3: 37964401-37964401 on Build GRCh38
rs568255452 Chr.3: 37964397-37964397 on Build GRCh38
rs1014564934 Chr.3: 37964408-37964408 on Build GRCh38
rs766233942 Chr.3: 37964561-37964561 on Build GRCh38
rs747059151 Chr.3: 37964530-37964530 on Build GRCh38
rs533992301 Chr.3: 37964399-37964399 on Build GRCh38
rs1023801167 Chr.3: 37964466-37964466 on Build GRCh38

View Full Product Details