Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15922263-15922530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F11
Entrez Gene ID
57834
Gene Name
cytochrome P450 family 4 subfamily F member 11
Gene Aliases
CYPIVF11
UniGene
Hs.187393
Gene Chromosome Location
Chr.19: 15912370-15934866 on Build GRCh38
Transcripts
NM_001128932.1 NM_021187.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5172007 c.985+9C>A p.? Chr.19: 15922355-15922355 on Build GRCh38
6150236 c.919-2A>G p.? Chr.19: 15922432-15922432 on Build GRCh38
992376 c.958G>T p.A320S Chr.19: 15922391-15922391 on Build GRCh38
992377 c.930G>T p.G310G Chr.19: 15922419-15922419 on Build GRCh38
4514386 c.972C>T p.T324T Chr.19: 15922377-15922377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777311850 Chr.19: 15922408-15922408 on Build GRCh38
rs71334792 Chr.19: 15922339-15922339 on Build GRCh38
rs767635975 Chr.19: 15922379-15922379 on Build GRCh38
rs993423351 Chr.19: 15922458-15922458 on Build GRCh38
rs762681796 Chr.19: 15922478-15922478 on Build GRCh38
rs200033002 Chr.19: 15922406-15922406 on Build GRCh38
rs114405853 Chr.19: 15922313-15922313 on Build GRCh38
rs770377786 Chr.19: 15922333-15922333 on Build GRCh38
rs567253288 Chr.19: 15922396-15922396 on Build GRCh38
rs1022414466 Chr.19: 15922291-15922291 on Build GRCh38
rs115830803 Chr.19: 15922298-15922298 on Build GRCh38
rs372854467 Chr.19: 15922397-15922397 on Build GRCh38
rs762000245 Chr.19: 15922370-15922370 on Build GRCh38
rs1049048148 Chr.19: 15922483-15922483 on Build GRCh38
rs748534533 Chr.19: 15922319-15922319 on Build GRCh38
rs375686924 Chr.19: 15922443-15922443 on Build GRCh38
rs775198259 Chr.19: 15922341-15922341 on Build GRCh38
rs751786236 Chr.19: 15922357-15922357 on Build GRCh38
rs866887636 Chr.19: 15922362-15922362 on Build GRCh38
rs745361090 Chr.19: 15922448-15922448 on Build GRCh38
rs746604840 Chr.19: 15922412-15922412 on Build GRCh38
rs762783310 Chr.19: 15922351-15922351 on Build GRCh38
rs764257394 Chr.19: 15922355-15922355 on Build GRCh38
rs1040019449 Chr.19: 15922410-15922410 on Build GRCh38
rs369120848 Chr.19: 15922331-15922331 on Build GRCh38
rs770568173 Chr.19: 15922425-15922425 on Build GRCh38
rs551550232 Chr.19: 15922401-15922401 on Build GRCh38
rs769659699 Chr.19: 15922459-15922459 on Build GRCh38
rs1050561010 Chr.19: 15922307-15922307 on Build GRCh38
rs538845657 Chr.19: 15922470-15922470 on Build GRCh38
rs547102668 Chr.19: 15922340-15922340 on Build GRCh38
rs754195024 Chr.19: 15922405-15922405 on Build GRCh38
rs867517429 Chr.19: 15922454-15922454 on Build GRCh38
rs569719220 Chr.19: 15922436-15922436 on Build GRCh38

View Full Product Details