Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 102888508-102888781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
UniGene
Hs.523446
Gene Chromosome Location
Chr.1: 102876467-103108580 on Build GRCh38
Transcripts
NM_001190709.1 NM_001854.3 NM_080629.2 NM_080630.3 NR_134980.1 XM_017000334.1 XM_017000335.1 XM_017000337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5842904 c.4556G>A p.G1519D Chr.1: 102888757-102888757 on Build GRCh38
5842903 c.4555-1G>A p.? Chr.1: 102888631-102888631 on Build GRCh38
1953808 c.4569G>A p.Q1523Q Chr.1: 102888616-102888616 on Build GRCh38
310203 c.4553C>T p.T1518I Chr.1: 102888724-102888724 on Build GRCh38
4549010 c.4628G>A p.G1543E Chr.1: 102888593-102888593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886044244 Chr.1: 102888722-102888722 on Build GRCh38
rs201561225 Chr.1: 102888564-102888564 on Build GRCh38
rs1676486 Chr.1: 102888582-102888582 on Build GRCh38
rs146997967 Chr.1: 102888604-102888604 on Build GRCh38
rs937559629 Chr.1: 102888708-102888708 on Build GRCh38
rs745798375 Chr.1: 102888606-102888606 on Build GRCh38
rs140954784 Chr.1: 102888591-102888591 on Build GRCh38
rs747193455 Chr.1: 102888690-102888690 on Build GRCh38
rs769726607 Chr.1: 102888533-102888533 on Build GRCh38
rs372206712 Chr.1: 102888715-102888715 on Build GRCh38
rs199597559 Chr.1: 102888543-102888543 on Build GRCh38
rs17127203 Chr.1: 102888559-102888559 on Build GRCh38
rs775358668 Chr.1: 102888536-102888536 on Build GRCh38
rs199882962 Chr.1: 102888556-102888556 on Build GRCh38
rs578137786 Chr.1: 102888599-102888599 on Build GRCh38
rs768543378 Chr.1: 102888672-102888672 on Build GRCh38
rs373610359 Chr.1: 102888537-102888537 on Build GRCh38
rs946274863 Chr.1: 102888669-102888669 on Build GRCh38
rs545281542 Chr.1: 102888689-102888689 on Build GRCh38
rs769920499 Chr.1: 102888624-102888624 on Build GRCh38
rs200793857 Chr.1: 102888704-102888704 on Build GRCh38
rs749149323 Chr.1: 102888660-102888660 on Build GRCh38
rs575231157 Chr.1: 102888732-102888732 on Build GRCh38
rs370574925 Chr.1: 102888613-102888613 on Build GRCh38
rs913393153 Chr.1: 102888663-102888663 on Build GRCh38
rs375198724 Chr.1: 102888686-102888686 on Build GRCh38
rs755033653 Chr.1: 102888646-102888646 on Build GRCh38
rs770918141 Chr.1: 102888696-102888696 on Build GRCh38
rs139064549 Chr.1: 102888579-102888579 on Build GRCh38
rs775945644 Chr.1: 102888747-102888747 on Build GRCh38
rs760018604 Chr.1: 102888570-102888570 on Build GRCh38
rs986809598 Chr.1: 102888583-102888583 on Build GRCh38
rs772483529 Chr.1: 102888541-102888541 on Build GRCh38
rs780129997 Chr.1: 102888625-102888625 on Build GRCh38
rs369145108 Chr.1: 102888691-102888691 on Build GRCh38

View Full Product Details