Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22086334-22086589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42
Entrez Gene ID
998
Gene Name
cell division cycle 42
Gene Aliases
CDC42Hs, G25K, TKS
UniGene
Hs.467637
Gene Chromosome Location
Chr.1: 22052627-22092943 on Build GRCh38
Transcripts
NM_001039802.1 NM_001791.3 NM_044472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
679405 c.283G>C p.E95Q Chr.1: 22086543-22086543 on Build GRCh38
4842328 c.374C>T p.S125L Chr.1: 22086508-22086508 on Build GRCh38
7080040 c.218C>T p.P73L Chr.1: 22086478-22086478 on Build GRCh38
5414401 c.188A>T p.D63V Chr.1: 22086448-22086448 on Build GRCh38
4751154 c.335T>C p.L112P Chr.1: 22086469-22086469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs797044870 Chr.1: 22086456-22086456 on Build GRCh38
rs746328036 Chr.1: 22086392-22086393 on Build GRCh38
rs774950267 Chr.1: 22086443-22086443 on Build GRCh38
rs905370439 Chr.1: 22086440-22086440 on Build GRCh38
rs761021558 Chr.1: 22086515-22086515 on Build GRCh38
rs750018506 Chr.1: 22086557-22086557 on Build GRCh38
rs749439872 Chr.1: 22086434-22086434 on Build GRCh38
rs16826534 Chr.1: 22086536-22086536 on Build GRCh38
rs778139162 Chr.1: 22086433-22086433 on Build GRCh38
rs747953542 Chr.1: 22086397-22086397 on Build GRCh38
rs372264403 Chr.1: 22086489-22086489 on Build GRCh38
rs368234616 Chr.1: 22086467-22086467 on Build GRCh38
rs770183176 Chr.1: 22086407-22086407 on Build GRCh38
rs529807426 Chr.1: 22086491-22086491 on Build GRCh38
rs864309721 Chr.1: 22086451-22086451 on Build GRCh38
rs758860745 Chr.1: 22086423-22086428 on Build GRCh38
rs771080774 Chr.1: 22086435-22086435 on Build GRCh38
rs772606537 Chr.1: 22086485-22086485 on Build GRCh38

View Full Product Details