Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9704712-9704949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989414 c.115G>C p.V39L Chr.3: 9704754-9704754 on Build GRCh38
6484338 c.206G>A p.R69H Chr.3: 9704940-9704940 on Build GRCh38
4447832 c.201C>T p.F67F Chr.3: 9704935-9704935 on Build GRCh38
4989415 c.115G>C p.V39L Chr.3: 9704754-9704754 on Build GRCh38
6457395 c.192C>G p.N64K Chr.3: 9704926-9704926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758742071 Chr.3: 9704911-9704911 on Build GRCh38
rs772442478 Chr.3: 9704778-9704778 on Build GRCh38
rs758556277 Chr.3: 9704878-9704878 on Build GRCh38
rs772904348 Chr.3: 9704797-9704797 on Build GRCh38
rs371129996 Chr.3: 9704813-9704813 on Build GRCh38
rs767291517 Chr.3: 9704809-9704809 on Build GRCh38
rs188299449 Chr.3: 9704837-9704837 on Build GRCh38
rs201517941 Chr.3: 9704754-9704754 on Build GRCh38
rs879067297 Chr.3: 9704814-9704814 on Build GRCh38
rs762565204 Chr.3: 9704804-9704804 on Build GRCh38
rs374162689 Chr.3: 9704850-9704850 on Build GRCh38
rs747587773 Chr.3: 9704791-9704791 on Build GRCh38
rs944047031 Chr.3: 9704877-9704878 on Build GRCh38
rs747496028 Chr.3: 9704835-9704835 on Build GRCh38
rs758945240 Chr.3: 9704828-9704828 on Build GRCh38
rs200930835 Chr.3: 9704773-9704773 on Build GRCh38
rs752128861 Chr.3: 9704920-9704920 on Build GRCh38
rs777917217 Chr.3: 9704832-9704832 on Build GRCh38
rs912607096 Chr.3: 9704868-9704868 on Build GRCh38
rs199691130 Chr.3: 9704818-9704818 on Build GRCh38
rs756409834 Chr.3: 9704741-9704741 on Build GRCh38
rs749002189 Chr.3: 9704841-9704841 on Build GRCh38
rs760658640 Chr.3: 9704881-9704881 on Build GRCh38
rs750827553 Chr.3: 9704910-9704910 on Build GRCh38
rs755935775 Chr.3: 9704808-9704809 on Build GRCh38
rs764246206 Chr.3: 9704817-9704817 on Build GRCh38
rs777278809 Chr.3: 9704929-9704929 on Build GRCh38
rs368749767 Chr.3: 9704820-9704820 on Build GRCh38
rs752768911 Chr.3: 9704730-9704730 on Build GRCh38
rs774005535 Chr.3: 9704806-9704806 on Build GRCh38
rs1053475581 Chr.3: 9704872-9704872 on Build GRCh38
rs757689742 Chr.3: 9704765-9704765 on Build GRCh38
rs1053600189 Chr.3: 9704734-9704734 on Build GRCh38
rs769462009 Chr.3: 9704794-9704794 on Build GRCh38
rs765305744 Chr.3: 9704905-9704905 on Build GRCh38
rs985506171 Chr.3: 9704851-9704851 on Build GRCh38
rs976441894 Chr.3: 9704803-9704803 on Build GRCh38
rs368322124 Chr.3: 9704807-9704807 on Build GRCh38
rs772069608 Chr.3: 9704877-9704877 on Build GRCh38
rs762105093 Chr.3: 9704900-9704900 on Build GRCh38
rs775616430 Chr.3: 9704880-9704880 on Build GRCh38
rs766703282 Chr.3: 9704913-9704913 on Build GRCh38
rs750693383 Chr.3: 9704826-9704826 on Build GRCh38
rs774203217 Chr.3: 9704863-9704863 on Build GRCh38
rs770828916 Chr.3: 9704805-9704805 on Build GRCh38
rs368004219 Chr.3: 9704882-9704882 on Build GRCh38
rs374289490 Chr.3: 9704780-9704780 on Build GRCh38
rs984752291 Chr.3: 9704740-9704740 on Build GRCh38
rs201599519 Chr.3: 9704866-9704866 on Build GRCh38
rs780495348 Chr.3: 9704834-9704834 on Build GRCh38
rs932357121 Chr.3: 9704812-9704812 on Build GRCh38
rs755467147 Chr.3: 9704926-9704926 on Build GRCh38
rs749181757 Chr.3: 9704835-9704840 on Build GRCh38
rs779388660 Chr.3: 9704824-9704824 on Build GRCh38
rs376552260 Chr.3: 9704760-9704760 on Build GRCh38
rs374524492 Chr.3: 9704819-9704819 on Build GRCh38
rs746452941 Chr.3: 9704776-9704776 on Build GRCh38

View Full Product Details