Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75089029-75089288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
UniGene
Hs.101302
Gene Chromosome Location
Chr.6: 75084326-75206262 on Build GRCh38
Transcripts
NM_004370.5 NM_080645.2 XM_011535434.1 XM_011535435.1 XM_011535436.2 XM_017010252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327181 c.8990G>A p.R2997Q Chr.6: 75089126-75089126 on Build GRCh38
3875468 c.9002G>C p.G3001A Chr.6: 75089114-75089114 on Build GRCh38
6297597 c.8989C>A p.R2997R Chr.6: 75089127-75089127 on Build GRCh38
5026589 c.8978C>T p.S2993F Chr.6: 75089138-75089138 on Build GRCh38
6632413 c.8960G>T p.G2987V Chr.6: 75089156-75089156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947201783 Chr.6: 75089188-75089188 on Build GRCh38
rs773428396 Chr.6: 75089074-75089074 on Build GRCh38
rs780326474 Chr.6: 75089144-75089144 on Build GRCh38
rs778475158 Chr.6: 75089186-75089205 on Build GRCh38
rs558202712 Chr.6: 75089078-75089078 on Build GRCh38
rs777153936 Chr.6: 75089179-75089179 on Build GRCh38
rs190501064 Chr.6: 75089146-75089146 on Build GRCh38
rs757075255 Chr.6: 75089127-75089127 on Build GRCh38
rs778638053 Chr.6: 75089126-75089126 on Build GRCh38
rs768999017 Chr.6: 75089169-75089169 on Build GRCh38
rs768754840 Chr.6: 75089062-75089062 on Build GRCh38
rs775198137 Chr.6: 75089215-75089215 on Build GRCh38
rs769877295 Chr.6: 75089203-75089203 on Build GRCh38
rs767271392 Chr.6: 75089091-75089091 on Build GRCh38
rs990502214 Chr.6: 75089246-75089246 on Build GRCh38
rs752674006 Chr.6: 75089095-75089095 on Build GRCh38
rs773411835 Chr.6: 75089204-75089204 on Build GRCh38
rs761904564 Chr.6: 75089065-75089065 on Build GRCh38
rs753345031 Chr.6: 75089111-75089111 on Build GRCh38
rs957302626 Chr.6: 75089220-75089220 on Build GRCh38
rs756855399 Chr.6: 75089112-75089112 on Build GRCh38
rs763149132 Chr.6: 75089208-75089208 on Build GRCh38
rs1006480073 Chr.6: 75089053-75089053 on Build GRCh38
rs748160231 Chr.6: 75089194-75089194 on Build GRCh38
rs34846477 Chr.6: 75089136-75089136 on Build GRCh38
rs765405314 Chr.6: 75089066-75089066 on Build GRCh38
rs756044625 Chr.6: 75089100-75089100 on Build GRCh38
rs1006254535 Chr.6: 75089073-75089073 on Build GRCh38
rs935359549 Chr.6: 75089231-75089231 on Build GRCh38
rs868456618 Chr.6: 75089121-75089121 on Build GRCh38
rs371038196 Chr.6: 75089063-75089063 on Build GRCh38
rs184663595 Chr.6: 75089110-75089110 on Build GRCh38
rs368624675 Chr.6: 75089057-75089057 on Build GRCh38
rs978662239 Chr.6: 75089196-75089196 on Build GRCh38
rs774597760 Chr.6: 75089058-75089059 on Build GRCh38
rs368067112 Chr.6: 75089197-75089197 on Build GRCh38
rs771868998 Chr.6: 75089214-75089214 on Build GRCh38
rs767363339 Chr.6: 75089131-75089131 on Build GRCh38
rs760537562 Chr.6: 75089224-75089224 on Build GRCh38
rs747539472 Chr.6: 75089058-75089058 on Build GRCh38

View Full Product Details