Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93061431-93061658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26C1
Entrez Gene ID
340665
Gene Name
cytochrome P450 family 26 subfamily C member 1
Gene Aliases
FFDD4
UniGene
Hs.369993
Gene Chromosome Location
Chr.10: 93061264-93068697 on Build GRCh38
Transcripts
NM_183374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5631240 c.171C>G p.P57P Chr.10: 93061434-93061434 on Build GRCh38
921527 c.177C>T p.F59F Chr.10: 93061440-93061440 on Build GRCh38
7255620 c.196delT p.L66fs*1 Chr.10: 93061459-93061459 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558650592 Chr.10: 93061570-93061570 on Build GRCh38
rs78631129 Chr.10: 93061627-93061627 on Build GRCh38
rs899091006 Chr.10: 93061512-93061512 on Build GRCh38
rs796462520 Chr.10: 93061627-93061632 on Build GRCh38
rs1055631926 Chr.10: 93061544-93061544 on Build GRCh38
rs1012273733 Chr.10: 93061565-93061565 on Build GRCh38
rs755174603 Chr.10: 93061448-93061448 on Build GRCh38
rs970679454 Chr.10: 93061591-93061591 on Build GRCh38
rs371261062 Chr.10: 93061471-93061471 on Build GRCh38
rs879431727 Chr.10: 93061627-93061627 on Build GRCh38
rs79885943 Chr.10: 93061630-93061633 on Build GRCh38
rs565550513 Chr.10: 93061626-93061627 on Build GRCh38
rs777714330 Chr.10: 93061492-93061492 on Build GRCh38
rs911544678 Chr.10: 93061472-93061472 on Build GRCh38
rs2422138 Chr.10: 93061617-93061617 on Build GRCh38
rs151114926 Chr.10: 93061522-93061522 on Build GRCh38
rs940360513 Chr.10: 93061485-93061485 on Build GRCh38
rs767361426 Chr.10: 93061454-93061454 on Build GRCh38
rs769882519 Chr.10: 93061623-93061625 on Build GRCh38
rs890479982 Chr.10: 93061535-93061535 on Build GRCh38
rs894334211 Chr.10: 93061545-93061545 on Build GRCh38
rs756016037 Chr.10: 93061488-93061488 on Build GRCh38
rs386746591 Chr.10: 93061627-93061632 on Build GRCh38
rs748776005 Chr.10: 93061503-93061503 on Build GRCh38
rs576446832 Chr.10: 93061630-93061632 on Build GRCh38
rs377621070 Chr.10: 93061600-93061600 on Build GRCh38
rs762244104 Chr.10: 93061628-93061629 on Build GRCh38
rs756828175 Chr.10: 93061517-93061517 on Build GRCh38
rs1037754860 Chr.10: 93061465-93061465 on Build GRCh38
rs4991544 Chr.10: 93061623-93061623 on Build GRCh38
rs1009131395 Chr.10: 93061555-93061555 on Build GRCh38
rs774230938 Chr.10: 93061628-93061632 on Build GRCh38
rs71736446 Chr.10: 93061629-93061632 on Build GRCh38
rs142915701 Chr.10: 93061627-93061631 on Build GRCh38
rs775594133 Chr.10: 93061629-93061633 on Build GRCh38
rs796804343 Chr.10: 93061626-93061627 on Build GRCh38
rs796822384 Chr.10: 93061632-93061632 on Build GRCh38
rs763160152 Chr.10: 93061631-93061634 on Build GRCh38
rs1025039769 Chr.10: 93061566-93061566 on Build GRCh38
rs377637297 Chr.10: 93061458-93061458 on Build GRCh38
rs1047770990 Chr.10: 93061531-93061531 on Build GRCh38

View Full Product Details