Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29231500-29231761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMID1
Entrez Gene ID
129080
Gene Name
EMI domain containing 1
Gene Aliases
EMI5, EMU1
UniGene
Hs.289106
Gene Chromosome Location
Chr.22: 29205912-29259597 on Build GRCh38
Transcripts
NM_001267895.1 NM_133455.3 XM_005261328.2 XM_005261329.2 XM_011529868.2 XM_011529869.2 XM_011529870.2 XM_011529871.2 XM_011529872.2 XM_011529873.2 XM_011529874.2 XM_011529875.2 XM_011529876.1 XM_017028589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1415469 c.587-2A>G p.? Chr.22: 29231591-29231591 on Build GRCh38
6526460 c.660G>A p.G220G Chr.22: 29231666-29231666 on Build GRCh38
225810 c.614G>A p.G205E Chr.22: 29231620-29231620 on Build GRCh38
1033116 c.639C>T p.A213A Chr.22: 29231645-29231645 on Build GRCh38
6359370 c.676+1G>C p.? Chr.22: 29231683-29231683 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749849046 Chr.22: 29231627-29231627 on Build GRCh38
rs369751814 Chr.22: 29231590-29231590 on Build GRCh38
rs368948271 Chr.22: 29231682-29231682 on Build GRCh38
rs908393305 Chr.22: 29231544-29231544 on Build GRCh38
rs766639012 Chr.22: 29231628-29231628 on Build GRCh38
rs201840101 Chr.22: 29231553-29231553 on Build GRCh38
rs112034362 Chr.22: 29231547-29231547 on Build GRCh38
rs573279440 Chr.22: 29231619-29231619 on Build GRCh38
rs537945996 Chr.22: 29231564-29231564 on Build GRCh38
rs555724508 Chr.22: 29231693-29231693 on Build GRCh38
rs748587620 Chr.22: 29231524-29231524 on Build GRCh38
rs895079766 Chr.22: 29231646-29231646 on Build GRCh38
rs752750191 Chr.22: 29231653-29231653 on Build GRCh38
rs538294511 Chr.22: 29231536-29231536 on Build GRCh38
rs145399834 Chr.22: 29231543-29231543 on Build GRCh38
rs78400471 Chr.22: 29231588-29231588 on Build GRCh38
rs567207310 Chr.22: 29231563-29231563 on Build GRCh38
rs905080934 Chr.22: 29231577-29231577 on Build GRCh38
rs1041714458 Chr.22: 29231565-29231577 on Build GRCh38
rs931097435 Chr.22: 29231534-29231534 on Build GRCh38
rs745473036 Chr.22: 29231703-29231703 on Build GRCh38
rs111764381 Chr.22: 29231522-29231522 on Build GRCh38
rs919586073 Chr.22: 29231523-29231523 on Build GRCh38
rs755633699 Chr.22: 29231581-29231581 on Build GRCh38
rs763038169 Chr.22: 29231601-29231601 on Build GRCh38
rs747462539 Chr.22: 29231677-29231677 on Build GRCh38
rs187827690 Chr.22: 29231552-29231552 on Build GRCh38
rs372860826 Chr.22: 29231618-29231618 on Build GRCh38
rs1041280689 Chr.22: 29231575-29231575 on Build GRCh38
rs966568366 Chr.22: 29231734-29231734 on Build GRCh38
rs767080669 Chr.22: 29231602-29231602 on Build GRCh38
rs758942795 Chr.22: 29231656-29231656 on Build GRCh38
rs769551739 Chr.22: 29231707-29231707 on Build GRCh38
rs975454710 Chr.22: 29231520-29231520 on Build GRCh38
rs753993763 Chr.22: 29231637-29231637 on Build GRCh38
rs550924502 Chr.22: 29231566-29231566 on Build GRCh38
rs977865304 Chr.22: 29231521-29231521 on Build GRCh38
rs374821978 Chr.22: 29231672-29231672 on Build GRCh38
rs534142288 Chr.22: 29231692-29231692 on Build GRCh38
rs760696015 Chr.22: 29231645-29231645 on Build GRCh38
rs755080930 Chr.22: 29231638-29231638 on Build GRCh38
rs200595514 Chr.22: 29231578-29231578 on Build GRCh38

View Full Product Details