Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35636282-35636543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2B1
Entrez Gene ID
163
Gene Name
adaptor related protein complex 2 beta 1 subunit
Gene Aliases
ADTB2, AP105B, AP2-BETA, CLAPB1
UniGene
Hs.514819
Gene Chromosome Location
Chr.17: 35586899-35726417 on Build GRCh38
Transcripts
NM_001030006.1 NM_001282.2 XM_005257937.3 XM_005257938.2 XM_005257941.2 XM_011524448.1 XM_011524449.2 XM_011524450.1 XM_011524451.1 XM_011524452.1 XM_011524453.1 XM_011524454.1 XM_011524455.2 XM_017024284.1 XM_017024285.1 XM_017024286.1 XM_017024287.1

SNP Detail

SNP ID Chromosome Location
rs777523979 Chr.17: 35636318-35636318 on Build GRCh38
rs374583609 Chr.17: 35636504-35636504 on Build GRCh38
rs746171249 Chr.17: 35636479-35636480 on Build GRCh38
rs553010896 Chr.17: 35636367-35636367 on Build GRCh38
rs760951186 Chr.17: 35636502-35636502 on Build GRCh38
rs780964830 Chr.17: 35636387-35636387 on Build GRCh38
rs367846442 Chr.17: 35636397-35636397 on Build GRCh38
rs370429115 Chr.17: 35636481-35636481 on Build GRCh38
rs755678145 Chr.17: 35636305-35636305 on Build GRCh38
rs545247814 Chr.17: 35636479-35636479 on Build GRCh38
rs912409539 Chr.17: 35636511-35636511 on Build GRCh38
rs746543051 Chr.17: 35636338-35636338 on Build GRCh38
rs147370808 Chr.17: 35636421-35636421 on Build GRCh38
rs779337192 Chr.17: 35636308-35636309 on Build GRCh38
rs975177290 Chr.17: 35636333-35636333 on Build GRCh38
rs769169713 Chr.17: 35636456-35636456 on Build GRCh38
rs932574938 Chr.17: 35636396-35636396 on Build GRCh38
rs749611432 Chr.17: 35636451-35636451 on Build GRCh38
rs774658819 Chr.17: 35636467-35636467 on Build GRCh38
rs780364265 Chr.17: 35636430-35636430 on Build GRCh38
rs912726039 Chr.17: 35636434-35636434 on Build GRCh38
rs773535938 Chr.17: 35636498-35636498 on Build GRCh38

View Full Product Details