Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133574044-133574293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDV3
Entrez Gene ID
55573
Gene Name
CDV3 homolog
Gene Aliases
H41
UniGene
Hs.518265
Gene Chromosome Location
Chr.3: 133573563-133590274 on Build GRCh38
Transcripts
NM_001134422.1 NM_001282762.1 NM_017548.4 XM_005247588.4 XM_005247589.4 XM_005247590.4 XM_005247591.4 XM_011512961.2 XM_017006808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612007 c.32A>C p.N11T Chr.3: 133574076-133574076 on Build GRCh38
5863548 c.52_54delAAG p.K22delK Chr.3: 133574096-133574098 on Build GRCh38
6414252 c.86C>T p.A29V Chr.3: 133574130-133574130 on Build GRCh38
6414253 c.86C>T p.A29V Chr.3: 133574130-133574130 on Build GRCh38
6409210 c.62A>T p.K21M Chr.3: 133574106-133574106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971557183 Chr.3: 133574144-133574144 on Build GRCh38
rs866902577 Chr.3: 133574086-133574086 on Build GRCh38
rs762476551 Chr.3: 133574107-133574107 on Build GRCh38
rs982469098 Chr.3: 133574161-133574161 on Build GRCh38
rs754534420 Chr.3: 133574132-133574132 on Build GRCh38
rs941107927 Chr.3: 133574194-133574194 on Build GRCh38
rs1037743592 Chr.3: 133574221-133574221 on Build GRCh38
rs1055548074 Chr.3: 133574191-133574191 on Build GRCh38
rs777615445 Chr.3: 133574187-133574187 on Build GRCh38
rs899637715 Chr.3: 133574205-133574205 on Build GRCh38
rs755927202 Chr.3: 133574178-133574178 on Build GRCh38
rs750691426 Chr.3: 133574138-133574146 on Build GRCh38
rs933818165 Chr.3: 133574206-133574206 on Build GRCh38
rs938444338 Chr.3: 133574182-133574182 on Build GRCh38
rs546879668 Chr.3: 133574143-133574143 on Build GRCh38
rs929607476 Chr.3: 133574151-133574151 on Build GRCh38
rs765955780 Chr.3: 133574123-133574123 on Build GRCh38
rs915603108 Chr.3: 133574199-133574199 on Build GRCh38
rs775679267 Chr.3: 133574062-133574062 on Build GRCh38
rs754381748 Chr.3: 133574104-133574104 on Build GRCh38
rs752364942 Chr.3: 133574162-133574162 on Build GRCh38
rs919518602 Chr.3: 133574148-133574148 on Build GRCh38
rs903098957 Chr.3: 133574272-133574272 on Build GRCh38
rs764449437 Chr.3: 133574098-133574098 on Build GRCh38
rs909574845 Chr.3: 133574189-133574191 on Build GRCh38
rs868410280 Chr.3: 133574077-133574077 on Build GRCh38
rs751160416 Chr.3: 133574128-133574128 on Build GRCh38
rs866240834 Chr.3: 133574080-133574080 on Build GRCh38
rs193280445 Chr.3: 133574168-133574168 on Build GRCh38
rs780926253 Chr.3: 133574152-133574152 on Build GRCh38
rs941663002 Chr.3: 133574203-133574203 on Build GRCh38
rs761107768 Chr.3: 133574068-133574068 on Build GRCh38
rs1023116340 Chr.3: 133574135-133574143 on Build GRCh38
rs1050956544 Chr.3: 133574233-133574233 on Build GRCh38
rs981630703 Chr.3: 133574147-133574147 on Build GRCh38
rs779337478 Chr.3: 133574096-133574098 on Build GRCh38

View Full Product Details