Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112679419-112679680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11AUN
Entrez Gene ID
400165
Gene Name
ATP11A upstream neighbor
Gene Aliases
C13orf35, SMABLO1
UniGene
Hs.591213
Gene Chromosome Location
Chr.13: 112647044-112684497 on Build GRCh38
Transcripts
NM_207440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6521247 c.222G>A p.R74R Chr.13: 112679601-112679601 on Build GRCh38
6217777 c.159G>A p.P53P Chr.13: 112679538-112679538 on Build GRCh38
6463071 c.134G>A p.R45H Chr.13: 112679513-112679513 on Build GRCh38
7185517 c.233G>T p.G78V Chr.13: 112679612-112679612 on Build GRCh38
7174034 c.90C>G p.A30A Chr.13: 112679469-112679469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760024974 Chr.13: 112679610-112679610 on Build GRCh38
rs757840603 Chr.13: 112679658-112679658 on Build GRCh38
rs1022006151 Chr.13: 112679534-112679534 on Build GRCh38
rs761194667 Chr.13: 112679640-112679640 on Build GRCh38
rs141230308 Chr.13: 112679506-112679506 on Build GRCh38
rs369426969 Chr.13: 112679477-112679477 on Build GRCh38
rs564116015 Chr.13: 112679527-112679527 on Build GRCh38
rs200954527 Chr.13: 112679564-112679564 on Build GRCh38
rs746932458 Chr.13: 112679524-112679524 on Build GRCh38
rs200914561 Chr.13: 112679597-112679597 on Build GRCh38
rs267603760 Chr.13: 112679458-112679458 on Build GRCh38
rs778304928 Chr.13: 112679510-112679521 on Build GRCh38
rs868395798 Chr.13: 112679559-112679564 on Build GRCh38
rs896136324 Chr.13: 112679594-112679594 on Build GRCh38
rs556365416 Chr.13: 112679456-112679456 on Build GRCh38
rs373171726 Chr.13: 112679483-112679483 on Build GRCh38
rs934042463 Chr.13: 112679502-112679502 on Build GRCh38
rs756425294 Chr.13: 112679508-112679508 on Build GRCh38
rs941735164 Chr.13: 112679446-112679446 on Build GRCh38
rs1033557756 Chr.13: 112679576-112679576 on Build GRCh38
rs939950123 Chr.13: 112679530-112679530 on Build GRCh38
rs746886400 Chr.13: 112679563-112679563 on Build GRCh38
rs752041784 Chr.13: 112679470-112679470 on Build GRCh38
rs751191674 Chr.13: 112679660-112679660 on Build GRCh38
rs764666222 Chr.13: 112679645-112679645 on Build GRCh38
rs192007935 Chr.13: 112679560-112679560 on Build GRCh38
rs745471588 Chr.13: 112679509-112679509 on Build GRCh38
rs765512223 Chr.13: 112679460-112679460 on Build GRCh38
rs866895795 Chr.13: 112679580-112679580 on Build GRCh38
rs367674877 Chr.13: 112679659-112679659 on Build GRCh38
rs150373081 Chr.13: 112679469-112679469 on Build GRCh38
rs117237810 Chr.13: 112679479-112679479 on Build GRCh38
rs138886345 Chr.13: 112679596-112679596 on Build GRCh38
rs891830358 Chr.13: 112679500-112679500 on Build GRCh38
rs754314917 Chr.13: 112679647-112679647 on Build GRCh38
rs750628088 Chr.13: 112679463-112679463 on Build GRCh38
rs1038142484 Chr.13: 112679533-112679533 on Build GRCh38
rs753284014 Chr.13: 112679629-112679629 on Build GRCh38
rs200943748 Chr.13: 112679512-112679512 on Build GRCh38
rs769893730 Chr.13: 112679593-112679593 on Build GRCh38
rs977688628 Chr.13: 112679655-112679655 on Build GRCh38
rs373831748 Chr.13: 112679648-112679648 on Build GRCh38
rs761923080 Chr.13: 112679536-112679536 on Build GRCh38
rs778442948 Chr.13: 112679558-112679558 on Build GRCh38
rs767941308 Chr.13: 112679621-112679621 on Build GRCh38
rs114091399 Chr.13: 112679495-112679495 on Build GRCh38
rs201512112 Chr.13: 112679489-112679489 on Build GRCh38
rs748029542 Chr.13: 112679589-112679589 on Build GRCh38
rs774434934 Chr.13: 112679607-112679607 on Build GRCh38
rs959182914 Chr.13: 112679586-112679586 on Build GRCh38
rs773329231 Chr.13: 112679542-112679542 on Build GRCh38
rs116657983 Chr.13: 112679443-112679443 on Build GRCh38
rs745508072 Chr.13: 112679559-112679559 on Build GRCh38
rs144955786 Chr.13: 112679535-112679535 on Build GRCh38
rs761865865 Chr.13: 112679459-112679459 on Build GRCh38
rs775288269 Chr.13: 112679515-112679515 on Build GRCh38
rs751897797 Chr.13: 112679544-112679544 on Build GRCh38
rs201364555 Chr.13: 112679556-112679556 on Build GRCh38
rs1024863286 Chr.13: 112679624-112679624 on Build GRCh38
rs199805592 Chr.13: 112679467-112679467 on Build GRCh38
rs755384113 Chr.13: 112679553-112679553 on Build GRCh38
rs765280632 Chr.13: 112679537-112679537 on Build GRCh38
rs758739806 Chr.13: 112679468-112679468 on Build GRCh38
rs768208765 Chr.13: 112679555-112679555 on Build GRCh38
rs763196221 Chr.13: 112679543-112679543 on Build GRCh38

View Full Product Details