Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23665499-23665766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN5
Entrez Gene ID
84516
Gene Name
dynactin subunit 5
Gene Aliases
-
UniGene
Hs.435941
Gene Chromosome Location
Chr.16: 23641366-23676207 on Build GRCh38
Transcripts
NM_001199743.1 NM_032486.3 NR_037573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507845 c.435C>A p.V145V Chr.16: 23665712-23665712 on Build GRCh38
4415154 c.408A>G p.P136P Chr.16: 23665685-23665685 on Build GRCh38
3772172 c.451+3A>T p.? Chr.16: 23665731-23665731 on Build GRCh38
968827 c.377G>T p.C126F Chr.16: 23665654-23665654 on Build GRCh38
4578837 c.363G>A p.V121V Chr.16: 23665640-23665640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022458782 Chr.16: 23665574-23665574 on Build GRCh38
rs775520489 Chr.16: 23665581-23665582 on Build GRCh38
rs905716849 Chr.16: 23665674-23665674 on Build GRCh38
rs776316761 Chr.16: 23665676-23665676 on Build GRCh38
rs753941051 Chr.16: 23665632-23665632 on Build GRCh38
rs149736104 Chr.16: 23665606-23665606 on Build GRCh38
rs35586 Chr.16: 23665685-23665685 on Build GRCh38
rs984934010 Chr.16: 23665703-23665703 on Build GRCh38
rs747389991 Chr.16: 23665669-23665669 on Build GRCh38
rs139923573 Chr.16: 23665527-23665527 on Build GRCh38
rs74012311 Chr.16: 23665554-23665554 on Build GRCh38
rs764107624 Chr.16: 23665728-23665728 on Build GRCh38
rs758623464 Chr.16: 23665655-23665655 on Build GRCh38
rs760861694 Chr.16: 23665607-23665609 on Build GRCh38
rs975970960 Chr.16: 23665645-23665645 on Build GRCh38
rs776103510 Chr.16: 23665575-23665575 on Build GRCh38
rs762965805 Chr.16: 23665722-23665722 on Build GRCh38
rs780480309 Chr.16: 23665657-23665657 on Build GRCh38
rs749834966 Chr.16: 23665526-23665526 on Build GRCh38
rs373307086 Chr.16: 23665629-23665629 on Build GRCh38
rs752808362 Chr.16: 23665627-23665627 on Build GRCh38
rs754327294 Chr.16: 23665733-23665733 on Build GRCh38
rs376809946 Chr.16: 23665579-23665579 on Build GRCh38
rs891942923 Chr.16: 23665548-23665548 on Build GRCh38
rs757459361 Chr.16: 23665633-23665633 on Build GRCh38
rs762616489 Chr.16: 23665604-23665604 on Build GRCh38
rs148262708 Chr.16: 23665640-23665640 on Build GRCh38
rs769325076 Chr.16: 23665699-23665699 on Build GRCh38
rs189912297 Chr.16: 23665585-23665585 on Build GRCh38
rs764481027 Chr.16: 23665630-23665630 on Build GRCh38
rs768348316 Chr.16: 23665672-23665672 on Build GRCh38
rs577497074 Chr.16: 23665529-23665529 on Build GRCh38
rs1001350013 Chr.16: 23665684-23665684 on Build GRCh38
rs375319296 Chr.16: 23665716-23665716 on Build GRCh38
rs766201486 Chr.16: 23665609-23665609 on Build GRCh38
rs773067755 Chr.16: 23665714-23665714 on Build GRCh38
rs371924708 Chr.16: 23665583-23665583 on Build GRCh38
rs774195056 Chr.16: 23665725-23665725 on Build GRCh38
rs746414090 Chr.16: 23665643-23665643 on Build GRCh38
rs150716217 Chr.16: 23665724-23665724 on Build GRCh38

View Full Product Details