Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43823842-43824095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1408138 c.1284C>T p.G428G Chr.2: 43823953-43823953 on Build GRCh38
4658481 c.1321C>A p.L441M Chr.2: 43823916-43823916 on Build GRCh38
5549609 c.1285G>A p.A429T Chr.2: 43823952-43823952 on Build GRCh38
3799022 c.1221C>G p.V407V Chr.2: 43824016-43824016 on Build GRCh38
1243058 c.1257C>T p.R419R Chr.2: 43823980-43823980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115590670 Chr.2: 43824009-43824009 on Build GRCh38
rs367702525 Chr.2: 43823867-43823867 on Build GRCh38
rs758152766 Chr.2: 43824022-43824022 on Build GRCh38
rs143680435 Chr.2: 43824014-43824014 on Build GRCh38
rs562178667 Chr.2: 43823915-43823915 on Build GRCh38
rs773653057 Chr.2: 43823900-43823900 on Build GRCh38
rs575266356 Chr.2: 43823926-43823926 on Build GRCh38
rs762490282 Chr.2: 43823912-43823912 on Build GRCh38
rs375364242 Chr.2: 43824020-43824020 on Build GRCh38
rs763515494 Chr.2: 43823996-43823996 on Build GRCh38
rs747481748 Chr.2: 43823979-43823979 on Build GRCh38
rs941620063 Chr.2: 43824043-43824043 on Build GRCh38
rs146801512 Chr.2: 43823986-43823986 on Build GRCh38
rs775019608 Chr.2: 43823907-43823907 on Build GRCh38
rs779125742 Chr.2: 43823952-43823952 on Build GRCh38
rs752962014 Chr.2: 43824021-43824021 on Build GRCh38
rs1044049813 Chr.2: 43823883-43823883 on Build GRCh38
rs772415178 Chr.2: 43823960-43823960 on Build GRCh38
rs761536339 Chr.2: 43824055-43824055 on Build GRCh38
rs867512029 Chr.2: 43824024-43824024 on Build GRCh38
rs897344189 Chr.2: 43824054-43824054 on Build GRCh38
rs146992652 Chr.2: 43824019-43824019 on Build GRCh38
rs868540610 Chr.2: 43824040-43824040 on Build GRCh38
rs143972075 Chr.2: 43823953-43823953 on Build GRCh38
rs119480069 Chr.2: 43824071-43824071 on Build GRCh38
rs907551949 Chr.2: 43823950-43823950 on Build GRCh38
rs373247682 Chr.2: 43823891-43823891 on Build GRCh38
rs773996683 Chr.2: 43823998-43823998 on Build GRCh38
rs767390913 Chr.2: 43824007-43824007 on Build GRCh38
rs949728635 Chr.2: 43823877-43823877 on Build GRCh38
rs746374174 Chr.2: 43824032-43824032 on Build GRCh38
rs573502874 Chr.2: 43823878-43823878 on Build GRCh38
rs764721117 Chr.2: 43823902-43823902 on Build GRCh38
rs751504404 Chr.2: 43823897-43823898 on Build GRCh38
rs780744534 Chr.2: 43823911-43823914 on Build GRCh38
rs148997255 Chr.2: 43824066-43824066 on Build GRCh38
rs765168105 Chr.2: 43823944-43823944 on Build GRCh38
rs119479067 Chr.2: 43823981-43823981 on Build GRCh38
rs535926485 Chr.2: 43823962-43823962 on Build GRCh38
rs773630787 Chr.2: 43823978-43823978 on Build GRCh38
rs201552973 Chr.2: 43823925-43823925 on Build GRCh38
rs754893727 Chr.2: 43823898-43823913 on Build GRCh38
rs119479065 Chr.2: 43824015-43824015 on Build GRCh38
rs371928191 Chr.2: 43823886-43823886 on Build GRCh38
rs765896139 Chr.2: 43824028-43824028 on Build GRCh38
rs751198635 Chr.2: 43823924-43823924 on Build GRCh38
rs368994670 Chr.2: 43823966-43823966 on Build GRCh38
rs978671799 Chr.2: 43823980-43823980 on Build GRCh38
rs771802286 Chr.2: 43824057-43824057 on Build GRCh38
rs144893516 Chr.2: 43824058-43824058 on Build GRCh38
rs150002815 Chr.2: 43823917-43823917 on Build GRCh38
rs1044958111 Chr.2: 43823987-43823987 on Build GRCh38
rs76576389 Chr.2: 43823868-43823868 on Build GRCh38
rs369261220 Chr.2: 43823892-43823892 on Build GRCh38
rs768197228 Chr.2: 43824042-43824042 on Build GRCh38
rs755044431 Chr.2: 43823945-43823945 on Build GRCh38
rs766160470 Chr.2: 43823882-43823883 on Build GRCh38
rs761230267 Chr.2: 43823901-43823901 on Build GRCh38
rs200396635 Chr.2: 43824027-43824027 on Build GRCh38
rs759461453 Chr.2: 43824072-43824072 on Build GRCh38
rs756162765 Chr.2: 43823933-43823933 on Build GRCh38
rs781675137 Chr.2: 43824031-43824031 on Build GRCh38
rs771475759 Chr.2: 43823982-43823982 on Build GRCh38
rs773776535 Chr.2: 43824048-43824048 on Build GRCh38
rs747622691 Chr.2: 43823990-43823991 on Build GRCh38
rs761429153 Chr.2: 43824001-43824001 on Build GRCh38

View Full Product Details