Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13918881-13919152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438701 c.1081G>C p.V361L Chr.19: 13918974-13918974 on Build GRCh38
1390873 c.1169G>A p.R390Q Chr.19: 13919149-13919149 on Build GRCh38
4923348 c.1021G>T p.V341L Chr.19: 13918914-13918914 on Build GRCh38
3822084 c.1064G>A p.R355Q Chr.19: 13918957-13918957 on Build GRCh38
3403821 c.1025C>A p.P342H Chr.19: 13918918-13918918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760924270 Chr.19: 13918979-13918979 on Build GRCh38
rs202234854 Chr.19: 13919073-13919073 on Build GRCh38
rs772187124 Chr.19: 13919048-13919048 on Build GRCh38
rs745472910 Chr.19: 13919028-13919028 on Build GRCh38
rs756869849 Chr.19: 13919019-13919019 on Build GRCh38
rs776994005 Chr.19: 13919059-13919059 on Build GRCh38
rs768626420 Chr.19: 13919034-13919034 on Build GRCh38
rs369829647 Chr.19: 13918914-13918914 on Build GRCh38
rs768483916 Chr.19: 13918946-13918946 on Build GRCh38
rs1056103923 Chr.19: 13918905-13918905 on Build GRCh38
rs755444041 Chr.19: 13919004-13919004 on Build GRCh38
rs779633150 Chr.19: 13919085-13919085 on Build GRCh38
rs745631694 Chr.19: 13918927-13918927 on Build GRCh38
rs748029345 Chr.19: 13919126-13919126 on Build GRCh38
rs373278035 Chr.19: 13918938-13918938 on Build GRCh38
rs754767706 Chr.19: 13919087-13919087 on Build GRCh38
rs772023620 Chr.19: 13919127-13919127 on Build GRCh38
rs755126900 Chr.19: 13919049-13919050 on Build GRCh38
rs773205133 Chr.19: 13919132-13919132 on Build GRCh38
rs377677009 Chr.19: 13919091-13919091 on Build GRCh38
rs775171615 Chr.19: 13918942-13918942 on Build GRCh38
rs779297750 Chr.19: 13919009-13919009 on Build GRCh38
rs779831737 Chr.19: 13919033-13919033 on Build GRCh38
rs558746804 Chr.19: 13918920-13918920 on Build GRCh38
rs868611556 Chr.19: 13918926-13918926 on Build GRCh38
rs753308817 Chr.19: 13919062-13919062 on Build GRCh38
rs760908544 Chr.19: 13919055-13919055 on Build GRCh38
rs750684031 Chr.19: 13918974-13918974 on Build GRCh38
rs576388238 Chr.19: 13918945-13918945 on Build GRCh38
rs749958194 Chr.19: 13919082-13919082 on Build GRCh38
rs774357851 Chr.19: 13918965-13918965 on Build GRCh38
rs766712869 Chr.19: 13918984-13918984 on Build GRCh38
rs759001406 Chr.19: 13919071-13919071 on Build GRCh38
rs766511788 Chr.19: 13919058-13919058 on Build GRCh38
rs781060128 Chr.19: 13918924-13918924 on Build GRCh38
rs537546040 Chr.19: 13918904-13918904 on Build GRCh38
rs369429952 Chr.19: 13919016-13919016 on Build GRCh38
rs758639508 Chr.19: 13919099-13919100 on Build GRCh38
rs996720493 Chr.19: 13919041-13919041 on Build GRCh38
rs767721110 Chr.19: 13918973-13918973 on Build GRCh38
rs748358885 Chr.19: 13919095-13919095 on Build GRCh38
rs1028164325 Chr.19: 13919050-13919050 on Build GRCh38
rs201420492 Chr.19: 13918988-13918988 on Build GRCh38
rs774337212 Chr.19: 13919036-13919036 on Build GRCh38
rs780974151 Chr.19: 13919027-13919027 on Build GRCh38
rs780355458 Chr.19: 13919086-13919086 on Build GRCh38
rs761937379 Chr.19: 13918966-13918966 on Build GRCh38
rs773298854 Chr.19: 13919054-13919054 on Build GRCh38
rs769364533 Chr.19: 13919031-13919031 on Build GRCh38
rs887935645 Chr.19: 13918928-13918928 on Build GRCh38
rs778799696 Chr.19: 13919125-13919125 on Build GRCh38
rs779027616 Chr.19: 13919059-13919059 on Build GRCh38

View Full Product Details