Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127577081-127577352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf58
Entrez Gene ID
352999
Gene Name
chromosome 6 open reading frame 58
Gene Aliases
LEG1
UniGene
Hs.226268
Gene Chromosome Location
Chr.6: 127577174-127591818 on Build GRCh38
Transcripts
NM_001010905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3620214 c.13C>T p.P5S Chr.6: 127577198-127577198 on Build GRCh38
3828987 c.153T>G p.I51M Chr.6: 127577338-127577338 on Build GRCh38
3620216 c.98G>A p.W33* Chr.6: 127577283-127577283 on Build GRCh38
1727165 c.116A>T p.Q39L Chr.6: 127577301-127577301 on Build GRCh38
3620215 c.41C>T p.S14F Chr.6: 127577226-127577226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9388590 Chr.6: 127577160-127577160 on Build GRCh38
rs562773334 Chr.6: 127577298-127577298 on Build GRCh38
rs542531909 Chr.6: 127577277-127577277 on Build GRCh38
rs763194353 Chr.6: 127577223-127577223 on Build GRCh38
rs772270512 Chr.6: 127577264-127577264 on Build GRCh38
rs755390630 Chr.6: 127577280-127577280 on Build GRCh38
rs371496473 Chr.6: 127577140-127577140 on Build GRCh38
rs766129589 Chr.6: 127577145-127577145 on Build GRCh38
rs753323414 Chr.6: 127577268-127577268 on Build GRCh38
rs747400916 Chr.6: 127577174-127577174 on Build GRCh38
rs1051883873 Chr.6: 127577113-127577113 on Build GRCh38
rs748733636 Chr.6: 127577205-127577205 on Build GRCh38
rs200591747 Chr.6: 127577272-127577272 on Build GRCh38
rs774678779 Chr.6: 127577235-127577235 on Build GRCh38
rs781729230 Chr.6: 127577282-127577282 on Build GRCh38
rs780429482 Chr.6: 127577168-127577168 on Build GRCh38
rs979461480 Chr.6: 127577281-127577281 on Build GRCh38
rs769851918 Chr.6: 127577209-127577209 on Build GRCh38
rs778407925 Chr.6: 127577273-127577273 on Build GRCh38
rs1014232244 Chr.6: 127577190-127577190 on Build GRCh38
rs201520968 Chr.6: 127577246-127577246 on Build GRCh38
rs760591268 Chr.6: 127577263-127577264 on Build GRCh38
rs752668146 Chr.6: 127577158-127577158 on Build GRCh38
rs150182872 Chr.6: 127577229-127577229 on Build GRCh38
rs1002210126 Chr.6: 127577207-127577207 on Build GRCh38
rs747532724 Chr.6: 127577219-127577219 on Build GRCh38
rs970328244 Chr.6: 127577240-127577240 on Build GRCh38
rs763596407 Chr.6: 127577259-127577259 on Build GRCh38
rs910321008 Chr.6: 127577142-127577142 on Build GRCh38
rs777248798 Chr.6: 127577189-127577189 on Build GRCh38
rs376465453 Chr.6: 127577121-127577121 on Build GRCh38
rs986161003 Chr.6: 127577139-127577139 on Build GRCh38
rs752615153 Chr.6: 127577247-127577247 on Build GRCh38
rs756061790 Chr.6: 127577256-127577256 on Build GRCh38
rs76385986 Chr.6: 127577165-127577165 on Build GRCh38
rs542972140 Chr.6: 127577119-127577119 on Build GRCh38
rs866332392 Chr.6: 127577253-127577253 on Build GRCh38
rs372875940 Chr.6: 127577164-127577164 on Build GRCh38
rs1007920368 Chr.6: 127577126-127577126 on Build GRCh38
rs781480375 Chr.6: 127577237-127577237 on Build GRCh38
rs748596729 Chr.6: 127577285-127577285 on Build GRCh38
rs554902065 Chr.6: 127577152-127577152 on Build GRCh38
rs769191716 Chr.6: 127577182-127577182 on Build GRCh38
rs373250656 Chr.6: 127577302-127577302 on Build GRCh38
rs535962054 Chr.6: 127577279-127577279 on Build GRCh38
rs778210931 Chr.6: 127577181-127577181 on Build GRCh38
rs896580457 Chr.6: 127577233-127577233 on Build GRCh38
rs767416828 Chr.6: 127577155-127577155 on Build GRCh38
rs756868272 Chr.6: 127577166-127577166 on Build GRCh38
rs777203492 Chr.6: 127577161-127577161 on Build GRCh38
rs376528445 Chr.6: 127577143-127577143 on Build GRCh38
rs773169017 Chr.6: 127577213-127577213 on Build GRCh38
rs779980609 Chr.6: 127577151-127577151 on Build GRCh38
rs745556346 Chr.6: 127577275-127577275 on Build GRCh38
rs879190438 Chr.6: 127577125-127577125 on Build GRCh38

View Full Product Details