Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154276664-154276889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227792 c.2799G>T p.W933C Chr.3: 154276789-154276789 on Build GRCh38
3365050 c.2657A>G p.Y886C Chr.3: 154276889-154276889 on Build GRCh38
4677406 c.2697delT p.F899fs*18 Chr.3: 154276849-154276849 on Build GRCh38
387380 c.2754C>A p.S918S Chr.3: 154276834-154276834 on Build GRCh38
4677404 c.2727C>T p.N909N Chr.3: 154276819-154276819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758737721 Chr.3: 154276792-154276792 on Build GRCh38
rs781438310 Chr.3: 154276811-154276811 on Build GRCh38
rs760190079 Chr.3: 154276739-154276739 on Build GRCh38
rs377323319 Chr.3: 154276819-154276819 on Build GRCh38
rs750771753 Chr.3: 154276790-154276790 on Build GRCh38
rs757479472 Chr.3: 154276860-154276860 on Build GRCh38
rs753427207 Chr.3: 154276756-154276756 on Build GRCh38
rs866726380 Chr.3: 154276830-154276830 on Build GRCh38
rs1016722808 Chr.3: 154276861-154276861 on Build GRCh38
rs142222613 Chr.3: 154276758-154276758 on Build GRCh38
rs759342254 Chr.3: 154276858-154276858 on Build GRCh38
rs752614061 Chr.3: 154276722-154276722 on Build GRCh38
rs768734415 Chr.3: 154276801-154276801 on Build GRCh38
rs776952795 Chr.3: 154276696-154276696 on Build GRCh38
rs762149375 Chr.3: 154276702-154276702 on Build GRCh38
rs9289943 Chr.3: 154276714-154276714 on Build GRCh38
rs756907366 Chr.3: 154276757-154276757 on Build GRCh38
rs374164936 Chr.3: 154276798-154276798 on Build GRCh38
rs371212695 Chr.3: 154276717-154276717 on Build GRCh38
rs151188512 Chr.3: 154276818-154276818 on Build GRCh38
rs386667254 Chr.3: 154276713-154276714 on Build GRCh38
rs200951715 Chr.3: 154276713-154276714 on Build GRCh38
rs759318435 Chr.3: 154276715-154276715 on Build GRCh38
rs771792767 Chr.3: 154276867-154276867 on Build GRCh38
rs998150992 Chr.3: 154276817-154276817 on Build GRCh38
rs777290837 Chr.3: 154276712-154276713 on Build GRCh38
rs902108724 Chr.3: 154276812-154276812 on Build GRCh38
rs138447229 Chr.3: 154276831-154276831 on Build GRCh38
rs748759274 Chr.3: 154276714-154276715 on Build GRCh38
rs355787 Chr.3: 154276713-154276713 on Build GRCh38
rs997410014 Chr.3: 154276742-154276742 on Build GRCh38
rs550827541 Chr.3: 154276741-154276741 on Build GRCh38

View Full Product Details