Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3523097-3523358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUAP1
Entrez Gene ID
23059
Gene Name
clusterin associated protein 1
Gene Aliases
CFAP22, FAP22, IFT38
UniGene
Hs.155995
Gene Chromosome Location
Chr.16: 3495562-3539048 on Build GRCh38
Transcripts
NM_015041.2 NM_024793.2 XM_006720867.3 XM_017023068.1 XM_017023069.1 XM_017023070.1 XM_017023071.1 XM_017023072.1 XM_017023073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6345127 c.780G>T p.L260L Chr.16: 3523224-3523224 on Build GRCh38
81031 c.771C>T p.D257D Chr.16: 3523215-3523215 on Build GRCh38
3948537 c.714-1G>A p.? Chr.16: 3523157-3523157 on Build GRCh38
6242649 c.815A>T p.Q272L Chr.16: 3523259-3523259 on Build GRCh38
6242650 c.92A>T p.Q31L Chr.16: 3523259-3523259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140813134 Chr.16: 3523328-3523328 on Build GRCh38
rs201028819 Chr.16: 3523131-3523131 on Build GRCh38
rs746068967 Chr.16: 3523307-3523307 on Build GRCh38
rs150154906 Chr.16: 3523209-3523209 on Build GRCh38
rs780823045 Chr.16: 3523273-3523273 on Build GRCh38
rs774621493 Chr.16: 3523148-3523148 on Build GRCh38
rs376920547 Chr.16: 3523306-3523306 on Build GRCh38
rs931955132 Chr.16: 3523142-3523142 on Build GRCh38
rs367831267 Chr.16: 3523284-3523284 on Build GRCh38
rs764693042 Chr.16: 3523224-3523224 on Build GRCh38
rs371974141 Chr.16: 3523304-3523304 on Build GRCh38
rs760747655 Chr.16: 3523315-3523315 on Build GRCh38
rs770758990 Chr.16: 3523305-3523305 on Build GRCh38
rs759823647 Chr.16: 3523176-3523176 on Build GRCh38
rs747678590 Chr.16: 3523280-3523280 on Build GRCh38
rs369835326 Chr.16: 3523138-3523138 on Build GRCh38
rs775508517 Chr.16: 3523313-3523313 on Build GRCh38
rs752823842 Chr.16: 3523215-3523215 on Build GRCh38
rs117478202 Chr.16: 3523217-3523217 on Build GRCh38
rs779201447 Chr.16: 3523253-3523253 on Build GRCh38
rs73503362 Chr.16: 3523248-3523248 on Build GRCh38
rs772305351 Chr.16: 3523310-3523310 on Build GRCh38
rs949081965 Chr.16: 3523325-3523325 on Build GRCh38
rs141573014 Chr.16: 3523299-3523299 on Build GRCh38
rs754082925 Chr.16: 3523240-3523240 on Build GRCh38
rs762814741 Chr.16: 3523126-3523126 on Build GRCh38
rs754714899 Chr.16: 3523268-3523268 on Build GRCh38
rs979752707 Chr.16: 3523316-3523316 on Build GRCh38
rs777180027 Chr.16: 3523324-3523324 on Build GRCh38
rs529162541 Chr.16: 3523162-3523162 on Build GRCh38
rs200622540 Chr.16: 3523137-3523137 on Build GRCh38
rs764449609 Chr.16: 3523319-3523319 on Build GRCh38
rs976073678 Chr.16: 3523127-3523127 on Build GRCh38
rs1035714055 Chr.16: 3523242-3523242 on Build GRCh38
rs751218423 Chr.16: 3523261-3523261 on Build GRCh38
rs769791977 Chr.16: 3523293-3523293 on Build GRCh38

View Full Product Details