Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78146758-78147025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf99
Entrez Gene ID
100141515
Gene Name
chromosome 17 open reading frame 99
Gene Aliases
UNQ464
UniGene
Hs.633034
Gene Chromosome Location
Chr.17: 78146035-78166283 on Build GRCh38
Transcripts
NM_001163075.1 XM_011524152.2 XM_017023997.1 XM_017023998.1 XM_017023999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985035 c.60A>G p.A20A Chr.17: 78146901-78146901 on Build GRCh38
5961591 c.61C>T p.R21W Chr.17: 78146902-78146902 on Build GRCh38
4130698 c.61C>A p.R21R Chr.17: 78146902-78146902 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755006670 Chr.17: 78146899-78146899 on Build GRCh38
rs750674033 Chr.17: 78146859-78146859 on Build GRCh38
rs7217374 Chr.17: 78146928-78146928 on Build GRCh38
rs986688785 Chr.17: 78146796-78146796 on Build GRCh38
rs1027910222 Chr.17: 78146786-78146786 on Build GRCh38
rs140831574 Chr.17: 78146861-78146861 on Build GRCh38
rs945632017 Chr.17: 78146978-78146978 on Build GRCh38
rs757408694 Chr.17: 78146814-78146814 on Build GRCh38
rs772962340 Chr.17: 78146933-78146933 on Build GRCh38
rs555959492 Chr.17: 78146815-78146815 on Build GRCh38
rs927103848 Chr.17: 78146889-78146889 on Build GRCh38
rs748974846 Chr.17: 78146947-78146947 on Build GRCh38
rs896639013 Chr.17: 78146974-78146974 on Build GRCh38
rs1049973714 Chr.17: 78146976-78146976 on Build GRCh38
rs778866896 Chr.17: 78146780-78146780 on Build GRCh38
rs553827043 Chr.17: 78146986-78146986 on Build GRCh38
rs144603878 Chr.17: 78146903-78146903 on Build GRCh38
rs938423806 Chr.17: 78146912-78146912 on Build GRCh38
rs765386971 Chr.17: 78146822-78146822 on Build GRCh38
rs954014906 Chr.17: 78146789-78146789 on Build GRCh38
rs932909436 Chr.17: 78146955-78146955 on Build GRCh38
rs770467911 Chr.17: 78146991-78146991 on Build GRCh38
rs370001743 Chr.17: 78146957-78146957 on Build GRCh38
rs569481927 Chr.17: 78146860-78146860 on Build GRCh38
rs980262122 Chr.17: 78146867-78146867 on Build GRCh38
rs540302911 Chr.17: 78146959-78146959 on Build GRCh38
rs1019858160 Chr.17: 78146797-78146797 on Build GRCh38
rs747632315 Chr.17: 78146924-78146924 on Build GRCh38
rs746887598 Chr.17: 78146894-78146894 on Build GRCh38
rs904689734 Chr.17: 78146998-78146998 on Build GRCh38
rs972606447 Chr.17: 78146828-78146828 on Build GRCh38

View Full Product Details