Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11313217-11313490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG7
Entrez Gene ID
10533
Gene Name
autophagy related 7
Gene Aliases
APG7-LIKE, APG7L, GSA7
UniGene
Hs.38032
Gene Chromosome Location
Chr.3: 11272324-11564704 on Build GRCh38
Transcripts
NM_001136031.2 NM_001144912.1 NM_006395.2 XM_006712931.3 XM_006712932.3 XM_006712933.3 XM_011533277.2 XM_011533278.2 XM_011533279.2 XM_011533280.2 XM_011533281.2 XM_011533282.2 XM_011533283.2 XM_011533284.2 XM_011533285.2 XM_011533286.2 XM_017005542.1 XM_017005543.1 XM_017005544.1 XM_017005545.1 XM_017005546.1 XM_017005547.1 XM_017005548.1 XM_017005549.1 XM_017005550.1 XM_017005551.1 XM_017005552.1 XM_017005553.1 XM_017005554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6882720 c.473G>A p.S158N Chr.3: 11313365-11313365 on Build GRCh38
365795 c.468A>G p.P156P Chr.3: 11313360-11313360 on Build GRCh38
479238 c.412-1G>T p.? Chr.3: 11313303-11313303 on Build GRCh38
419920 c.420G>C p.K140N Chr.3: 11313312-11313312 on Build GRCh38
5449767 c.512G>T p.R171M Chr.3: 11313404-11313404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377056082 Chr.3: 11313288-11313288 on Build GRCh38
rs755610196 Chr.3: 11313448-11313448 on Build GRCh38
rs113070394 Chr.3: 11313331-11313331 on Build GRCh38
rs770943091 Chr.3: 11313343-11313343 on Build GRCh38
rs745728273 Chr.3: 11313368-11313368 on Build GRCh38
rs553156697 Chr.3: 11313456-11313456 on Build GRCh38
rs747599089 Chr.3: 11313326-11313326 on Build GRCh38
rs763418077 Chr.3: 11313428-11313428 on Build GRCh38
rs150258952 Chr.3: 11313400-11313400 on Build GRCh38
rs375008041 Chr.3: 11313351-11313351 on Build GRCh38
rs143084842 Chr.3: 11313341-11313341 on Build GRCh38
rs755596978 Chr.3: 11313327-11313327 on Build GRCh38
rs768645298 Chr.3: 11313395-11313395 on Build GRCh38
rs200786439 Chr.3: 11313443-11313443 on Build GRCh38
rs530909987 Chr.3: 11313252-11313252 on Build GRCh38
rs140608347 Chr.3: 11313319-11313319 on Build GRCh38
rs746323066 Chr.3: 11313290-11313290 on Build GRCh38
rs755216792 Chr.3: 11313292-11313292 on Build GRCh38
rs754666048 Chr.3: 11313302-11313302 on Build GRCh38
rs772213768 Chr.3: 11313369-11313369 on Build GRCh38
rs549229057 Chr.3: 11313329-11313329 on Build GRCh38
rs201384830 Chr.3: 11313411-11313411 on Build GRCh38
rs778658863 Chr.3: 11313360-11313360 on Build GRCh38
rs754323858 Chr.3: 11313271-11313271 on Build GRCh38
rs760759434 Chr.3: 11313389-11313389 on Build GRCh38
rs921217047 Chr.3: 11313336-11313336 on Build GRCh38
rs764602356 Chr.3: 11313263-11313263 on Build GRCh38
rs1020397223 Chr.3: 11313384-11313384 on Build GRCh38
rs1008967884 Chr.3: 11313378-11313378 on Build GRCh38
rs372010423 Chr.3: 11313254-11313254 on Build GRCh38
rs775702529 Chr.3: 11313371-11313371 on Build GRCh38
rs750865245 Chr.3: 11313299-11313299 on Build GRCh38
rs908281343 Chr.3: 11313298-11313298 on Build GRCh38
rs982471363 Chr.3: 11313262-11313262 on Build GRCh38
rs1053223896 Chr.3: 11313413-11313413 on Build GRCh38
rs895898102 Chr.3: 11313430-11313430 on Build GRCh38
rs370094457 Chr.3: 11313423-11313423 on Build GRCh38
rs775541165 Chr.3: 11313253-11313253 on Build GRCh38
rs770308401 Chr.3: 11313374-11313374 on Build GRCh38
rs965211964 Chr.3: 11313324-11313324 on Build GRCh38
rs144786187 Chr.3: 11313405-11313405 on Build GRCh38
rs763673542 Chr.3: 11313277-11313277 on Build GRCh38
rs767224966 Chr.3: 11313435-11313435 on Build GRCh38

View Full Product Details