Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131084577-131084828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP36
Entrez Gene ID
158763
Gene Name
Rho GTPase activating protein 36
Gene Aliases
-
UniGene
Hs.22905
Gene Chromosome Location
Chr.X: 131058242-131089885 on Build GRCh38
Transcripts
NM_001282607.1 NM_144967.3 XM_006724729.1 XM_011531280.1

SNP Detail

SNP ID Chromosome Location
rs745425289 Chr.X: 131084644-131084644 on Build GRCh38
rs756999878 Chr.X: 131084607-131084607 on Build GRCh38
rs374674459 Chr.X: 131084769-131084769 on Build GRCh38
rs774848282 Chr.X: 131084726-131084726 on Build GRCh38
rs151036602 Chr.X: 131084640-131084640 on Build GRCh38
rs960206394 Chr.X: 131084771-131084771 on Build GRCh38
rs755037555 Chr.X: 131084748-131084748 on Build GRCh38
rs769255505 Chr.X: 131084664-131084664 on Build GRCh38
rs370750175 Chr.X: 131084634-131084634 on Build GRCh38
rs368655030 Chr.X: 131084606-131084606 on Build GRCh38
rs779030925 Chr.X: 131084767-131084768 on Build GRCh38
rs752848208 Chr.X: 131084768-131084768 on Build GRCh38
rs750411009 Chr.X: 131084602-131084602 on Build GRCh38
rs952393953 Chr.X: 131084633-131084633 on Build GRCh38
rs749738678 Chr.X: 131084702-131084702 on Build GRCh38
rs769016364 Chr.X: 131084711-131084711 on Build GRCh38
rs1055699359 Chr.X: 131084777-131084777 on Build GRCh38
rs780560921 Chr.X: 131084694-131084694 on Build GRCh38

View Full Product Details