Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35175570-35175837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREM
Entrez Gene ID
1390
Gene Name
cAMP responsive element modulator
Gene Aliases
CREM-2, ICER, hCREM-2
UniGene
Hs.200250
Gene Chromosome Location
Chr.10: 35126841-35212958 on Build GRCh38
Transcripts
NM_001267562.1 NM_001267563.1 NM_001267564.1 NM_001267565.1 NM_001267566.1 NM_001267567.1 NM_001881.3 NM_181571.2 NM_182769.2 NM_182770.2 NM_182771.1 NM_182772.1 NM_183011.1 NM_183012.1 NM_183013.2 NM_183060.2 NR_051973.1 NR_051974.1 NR_051975.1 XM_006717378.3 XM_006717379.3 XM_006717382.3 XM_006717383.3 XM_006717387.3 XM_011519316.2 XM_011519324.2 XM_011519325.2 XM_011519327.2 XM_011519330.2 XM_011519331.2 XM_011519332.2 XM_011519333.1 XM_011519335.2 XM_017015716.1 XM_017015717.1 XM_017015718.1 XM_017015719.1 XM_017015720.1 XM_017015721.1 XM_017015722.1 XM_017015723.1 XM_017015724.1 XM_017015725.1 XM_017015726.1 XM_017015727.1 XM_017015728.1 XM_017015729.1 XM_017015730.1 XM_017015731.1 XM_017015732.1 XM_017015733.1 XM_017015734.1 XM_017015735.1 XM_017015736.1 XM_017015737.1

SNP Detail

SNP ID Chromosome Location
rs773233312 Chr.10: 35175814-35175814 on Build GRCh38
rs929948288 Chr.10: 35175766-35175766 on Build GRCh38
rs770080709 Chr.10: 35175809-35175809 on Build GRCh38
rs897272870 Chr.10: 35175762-35175762 on Build GRCh38
rs958295181 Chr.10: 35175789-35175789 on Build GRCh38
rs112311636 Chr.10: 35175744-35175744 on Build GRCh38
rs761021435 Chr.10: 35175722-35175723 on Build GRCh38
rs138132743 Chr.10: 35175701-35175701 on Build GRCh38
rs141314816 Chr.10: 35175610-35175610 on Build GRCh38
rs781048644 Chr.10: 35175803-35175803 on Build GRCh38
rs767982309 Chr.10: 35175695-35175695 on Build GRCh38
rs752096148 Chr.10: 35175688-35175688 on Build GRCh38
rs546750933 Chr.10: 35175697-35175697 on Build GRCh38
rs111589791 Chr.10: 35175665-35175665 on Build GRCh38
rs759523738 Chr.10: 35175660-35175660 on Build GRCh38
rs780119724 Chr.10: 35175736-35175736 on Build GRCh38
rs41276100 Chr.10: 35175760-35175760 on Build GRCh38
rs928539952 Chr.10: 35175681-35175681 on Build GRCh38
rs898171926 Chr.10: 35175667-35175667 on Build GRCh38
rs754728642 Chr.10: 35175750-35175750 on Build GRCh38
rs570271926 Chr.10: 35175608-35175608 on Build GRCh38
rs766577328 Chr.10: 35175611-35175611 on Build GRCh38
rs370349059 Chr.10: 35175725-35175725 on Build GRCh38
rs377100626 Chr.10: 35175711-35175711 on Build GRCh38
rs763405299 Chr.10: 35175671-35175671 on Build GRCh38
rs759794387 Chr.10: 35175691-35175691 on Build GRCh38
rs749984746 Chr.10: 35175715-35175715 on Build GRCh38
rs372408025 Chr.10: 35175743-35175743 on Build GRCh38
rs368064158 Chr.10: 35175804-35175804 on Build GRCh38
rs981161933 Chr.10: 35175663-35175663 on Build GRCh38
rs942635424 Chr.10: 35175646-35175646 on Build GRCh38
rs777962211 Chr.10: 35175674-35175684 on Build GRCh38
rs758448596 Chr.10: 35175702-35175702 on Build GRCh38
rs765304676 Chr.10: 35175693-35175693 on Build GRCh38
rs374023956 Chr.10: 35175676-35175676 on Build GRCh38

View Full Product Details