Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27500827-27501100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX2
Entrez Gene ID
2053
Gene Name
epoxide hydrolase 2
Gene Aliases
CEH, SEH
UniGene
Hs.212088
Gene Chromosome Location
Chr.8: 27491002-27597559 on Build GRCh38
Transcripts
NM_001256482.1 NM_001256483.1 NM_001256484.1 NM_001979.5 XM_017013199.1 XM_017013200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
749942 c.184C>A p.Q62K Chr.8: 27501008-27501008 on Build GRCh38
136655 c.166G>A p.G56R Chr.8: 27500990-27500990 on Build GRCh38
1098744 c.138A>C p.P46P Chr.8: 27500962-27500962 on Build GRCh38
7146291 c.174C>A p.I58I Chr.8: 27500998-27500998 on Build GRCh38
4366885 c.139G>T p.E47* Chr.8: 27500963-27500963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766492336 Chr.8: 27501020-27501020 on Build GRCh38
rs762135838 Chr.8: 27500939-27500939 on Build GRCh38
rs888952470 Chr.8: 27500957-27500957 on Build GRCh38
rs1009719941 Chr.8: 27500990-27500990 on Build GRCh38
rs939536565 Chr.8: 27500905-27500905 on Build GRCh38
rs749370691 Chr.8: 27500935-27500935 on Build GRCh38
rs781604277 Chr.8: 27500993-27500993 on Build GRCh38
rs755352071 Chr.8: 27500908-27500908 on Build GRCh38
rs377050303 Chr.8: 27500931-27500931 on Build GRCh38
rs753014790 Chr.8: 27500994-27500994 on Build GRCh38
rs533460949 Chr.8: 27500998-27500998 on Build GRCh38
rs371491337 Chr.8: 27500872-27500872 on Build GRCh38
rs748687158 Chr.8: 27500929-27500929 on Build GRCh38
rs779884282 Chr.8: 27500896-27500896 on Build GRCh38
rs754657097 Chr.8: 27500992-27500992 on Build GRCh38
rs374262086 Chr.8: 27501060-27501060 on Build GRCh38
rs370170529 Chr.8: 27501016-27501016 on Build GRCh38
rs561926285 Chr.8: 27500939-27500939 on Build GRCh38
rs374362897 Chr.8: 27501027-27501027 on Build GRCh38
rs138702597 Chr.8: 27501012-27501012 on Build GRCh38
rs770976715 Chr.8: 27501021-27501021 on Build GRCh38
rs139234214 Chr.8: 27500978-27500978 on Build GRCh38
rs776423485 Chr.8: 27501046-27501046 on Build GRCh38
rs745900946 Chr.8: 27501044-27501044 on Build GRCh38
rs773263366 Chr.8: 27501062-27501062 on Build GRCh38
rs188742114 Chr.8: 27501000-27501000 on Build GRCh38
rs774577848 Chr.8: 27500940-27500940 on Build GRCh38
rs374549372 Chr.8: 27500909-27500909 on Build GRCh38
rs767630759 Chr.8: 27501031-27501032 on Build GRCh38
rs758007869 Chr.8: 27500983-27500983 on Build GRCh38
rs758058631 Chr.8: 27500880-27500880 on Build GRCh38
rs372114163 Chr.8: 27501049-27501049 on Build GRCh38
rs772199523 Chr.8: 27501034-27501034 on Build GRCh38
rs565274773 Chr.8: 27500877-27500877 on Build GRCh38
rs776349097 Chr.8: 27500973-27500973 on Build GRCh38
rs41507953 Chr.8: 27500988-27500988 on Build GRCh38
rs201108990 Chr.8: 27500932-27500932 on Build GRCh38
rs747331659 Chr.8: 27500902-27500902 on Build GRCh38
rs376799140 Chr.8: 27500930-27500930 on Build GRCh38
rs766040550 Chr.8: 27500985-27500985 on Build GRCh38
rs774099359 Chr.8: 27500971-27500971 on Build GRCh38
rs1022274678 Chr.8: 27501075-27501075 on Build GRCh38
rs763498934 Chr.8: 27500954-27500954 on Build GRCh38
rs367964364 Chr.8: 27501048-27501048 on Build GRCh38
rs72475803 Chr.8: 27500979-27500979 on Build GRCh38
rs373720159 Chr.8: 27501068-27501068 on Build GRCh38
rs919542858 Chr.8: 27500956-27500956 on Build GRCh38

View Full Product Details