Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135033943-135034192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_011516122.2 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925329 c.480-1G>A p.? Chr.7: 135034148-135034148 on Build GRCh38
5141579 c.505G>A p.V169I Chr.7: 135034174-135034174 on Build GRCh38
1448228 c.583G>A p.V195I Chr.7: 135034174-135034174 on Build GRCh38
5925331 c.480G>A p.V160V Chr.7: 135034149-135034149 on Build GRCh38
5794124 c.567C>T p.Y189Y Chr.7: 135034158-135034158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762990545 Chr.7: 135034116-135034116 on Build GRCh38
rs756148484 Chr.7: 135034147-135034147 on Build GRCh38
rs371154222 Chr.7: 135034158-135034158 on Build GRCh38
rs1039072895 Chr.7: 135034069-135034069 on Build GRCh38
rs901517221 Chr.7: 135034077-135034077 on Build GRCh38
rs1007285120 Chr.7: 135034063-135034063 on Build GRCh38
rs764050909 Chr.7: 135034131-135034131 on Build GRCh38
rs752416451 Chr.7: 135034120-135034121 on Build GRCh38
rs892415219 Chr.7: 135033978-135033978 on Build GRCh38
rs945207610 Chr.7: 135033980-135033980 on Build GRCh38
rs1003982656 Chr.7: 135034019-135034019 on Build GRCh38
rs533384728 Chr.7: 135034012-135034012 on Build GRCh38
rs991617023 Chr.7: 135034083-135034083 on Build GRCh38
rs926581325 Chr.7: 135034159-135034159 on Build GRCh38
rs1017075402 Chr.7: 135034074-135034074 on Build GRCh38
rs914633930 Chr.7: 135034162-135034162 on Build GRCh38
rs35722574 Chr.7: 135033977-135033978 on Build GRCh38
rs1004370041 Chr.7: 135034117-135034117 on Build GRCh38
rs191258986 Chr.7: 135034133-135034133 on Build GRCh38
rs934640750 Chr.7: 135034143-135034143 on Build GRCh38
rs767089975 Chr.7: 135034142-135034142 on Build GRCh38
rs775270273 Chr.7: 135034111-135034111 on Build GRCh38
rs981632223 Chr.7: 135034129-135034129 on Build GRCh38
rs569985457 Chr.7: 135034016-135034016 on Build GRCh38
rs898054749 Chr.7: 135034161-135034161 on Build GRCh38
rs997626340 Chr.7: 135034091-135034091 on Build GRCh38
rs924655606 Chr.7: 135034141-135034141 on Build GRCh38
rs560669502 Chr.7: 135034079-135034079 on Build GRCh38

View Full Product Details