Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41968168-41968439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 XM_006721701.3 XM_011524340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201631 c.185C>T p.S62L Chr.17: 41968309-41968309 on Build GRCh38
1325075 c.190G>C p.D64H Chr.17: 41968314-41968314 on Build GRCh38
4066482 c.79C>A p.L27I Chr.17: 41968203-41968203 on Build GRCh38
6230423 c.291C>T p.S97S Chr.17: 41968355-41968355 on Build GRCh38
6256289 c.316G>T p.D106Y Chr.17: 41968380-41968380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782195746 Chr.17: 41968274-41968274 on Build GRCh38
rs781840726 Chr.17: 41968404-41968404 on Build GRCh38
rs782058156 Chr.17: 41968352-41968352 on Build GRCh38
rs946697398 Chr.17: 41968301-41968301 on Build GRCh38
rs782430594 Chr.17: 41968354-41968354 on Build GRCh38
rs781970302 Chr.17: 41968287-41968287 on Build GRCh38
rs138346889 Chr.17: 41968232-41968232 on Build GRCh38
rs782349606 Chr.17: 41968348-41968348 on Build GRCh38
rs782619997 Chr.17: 41968334-41968334 on Build GRCh38
rs782683469 Chr.17: 41968316-41968316 on Build GRCh38
rs997851332 Chr.17: 41968344-41968344 on Build GRCh38
rs143924216 Chr.17: 41968322-41968322 on Build GRCh38
rs782128196 Chr.17: 41968366-41968366 on Build GRCh38
rs368980681 Chr.17: 41968269-41968269 on Build GRCh38
rs781968189 Chr.17: 41968193-41968193 on Build GRCh38
rs782564312 Chr.17: 41968223-41968223 on Build GRCh38
rs782088799 Chr.17: 41968197-41968197 on Build GRCh38
rs372379354 Chr.17: 41968199-41968199 on Build GRCh38
rs374975038 Chr.17: 41968342-41968342 on Build GRCh38
rs782433665 Chr.17: 41968310-41968310 on Build GRCh38
rs555951916 Chr.17: 41968355-41968355 on Build GRCh38
rs902752168 Chr.17: 41968311-41968311 on Build GRCh38
rs781801341 Chr.17: 41968205-41968205 on Build GRCh38
rs781943089 Chr.17: 41968349-41968349 on Build GRCh38
rs782779483 Chr.17: 41968391-41968391 on Build GRCh38
rs782599531 Chr.17: 41968273-41968273 on Build GRCh38
rs1043767606 Chr.17: 41968307-41968307 on Build GRCh38
rs782788519 Chr.17: 41968231-41968231 on Build GRCh38
rs902026818 Chr.17: 41968338-41968338 on Build GRCh38

View Full Product Details