Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22329266-22329535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMI1
Entrez Gene ID
648
Gene Name
BMI1 proto-oncogene, polycomb ring finger
Gene Aliases
FLVI2/BMI1, PCGF4, RNF51, flvi-2/bmi-1
UniGene
Hs.380403
Gene Chromosome Location
Chr.10: 22321210-22331485 on Build GRCh38
Transcripts
NM_005180.8 NM_001204062.1

Gene Symbol
COMMD3-BMI1
Entrez Gene ID
100532731
Gene Name
COMMD3-BMI1 readthrough
Gene Aliases
-
UniGene
Hs.380403
Gene Chromosome Location
Chr.10: 22316383-22331485 on Build GRCh38
Transcripts
NM_005180.8 NM_001204062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3436549 c.832C>T p.P278S Chr.10: 22329393-22329393 on Build GRCh38
5753450 c.930T>G p.N310K Chr.10: 22329491-22329491 on Build GRCh38
917041 c.938G>A p.R313Q Chr.10: 22329499-22329499 on Build GRCh38
917040 c.837G>A p.V279V Chr.10: 22329398-22329398 on Build GRCh38
917039 c.766G>A p.D256N Chr.10: 22329327-22329327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760461431 Chr.10: 22329357-22329357 on Build GRCh38
rs757966770 Chr.10: 22329498-22329498 on Build GRCh38
rs778305888 Chr.10: 22329379-22329379 on Build GRCh38
rs145613627 Chr.10: 22329418-22329418 on Build GRCh38
rs866781440 Chr.10: 22329454-22329454 on Build GRCh38
rs766114933 Chr.10: 22329480-22329480 on Build GRCh38
rs368232993 Chr.10: 22329415-22329415 on Build GRCh38
rs201024480 Chr.10: 22329490-22329490 on Build GRCh38
rs142604751 Chr.10: 22329495-22329495 on Build GRCh38
rs752226313 Chr.10: 22329385-22329385 on Build GRCh38
rs376521566 Chr.10: 22329387-22329387 on Build GRCh38
rs745326959 Chr.10: 22329353-22329354 on Build GRCh38
rs202165238 Chr.10: 22329298-22329298 on Build GRCh38
rs140326477 Chr.10: 22329505-22329505 on Build GRCh38
rs759158310 Chr.10: 22329491-22329491 on Build GRCh38
rs769364356 Chr.10: 22329482-22329483 on Build GRCh38
rs753450025 Chr.10: 22329367-22329367 on Build GRCh38
rs768622895 Chr.10: 22329296-22329296 on Build GRCh38
rs773125077 Chr.10: 22329474-22329474 on Build GRCh38
rs554358305 Chr.10: 22329464-22329464 on Build GRCh38
rs148911593 Chr.10: 22329435-22329435 on Build GRCh38
rs754615197 Chr.10: 22329373-22329373 on Build GRCh38
rs761853504 Chr.10: 22329465-22329465 on Build GRCh38
rs374810543 Chr.10: 22329499-22329499 on Build GRCh38
rs987052300 Chr.10: 22329506-22329506 on Build GRCh38
rs750261916 Chr.10: 22329337-22329337 on Build GRCh38
rs766117385 Chr.10: 22329365-22329365 on Build GRCh38
rs76089553 Chr.10: 22329496-22329496 on Build GRCh38
rs772066505 Chr.10: 22329471-22329471 on Build GRCh38
rs749504587 Chr.10: 22329440-22329440 on Build GRCh38
rs768875896 Chr.10: 22329461-22329461 on Build GRCh38
rs767421037 Chr.10: 22329335-22329335 on Build GRCh38
rs764909255 Chr.10: 22329493-22329493 on Build GRCh38
rs1009236602 Chr.10: 22329336-22329336 on Build GRCh38
rs780400625 Chr.10: 22329438-22329438 on Build GRCh38
rs760551695 Chr.10: 22329476-22329476 on Build GRCh38
rs761763487 Chr.10: 22329305-22329305 on Build GRCh38
rs763640328 Chr.10: 22329510-22329510 on Build GRCh38

View Full Product Details