Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57382546-57382785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CL1
Entrez Gene ID
6376
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
UniGene
Hs.531668
Gene Chromosome Location
Chr.16: 57372461-57385048 on Build GRCh38
Transcripts
NM_001304392.1 NM_002996.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903095 c.721C>T p.P241S Chr.16: 57382559-57382559 on Build GRCh38
971824 c.856G>A p.V286I Chr.16: 57382694-57382694 on Build GRCh38
4675687 c.788A>G p.E263G Chr.16: 57382626-57382626 on Build GRCh38
5549347 c.931G>T p.A311S Chr.16: 57382769-57382769 on Build GRCh38
6582251 c.934G>C p.E312Q Chr.16: 57382772-57382772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754502293 Chr.16: 57382709-57382709 on Build GRCh38
rs201467674 Chr.16: 57382653-57382653 on Build GRCh38
rs746808529 Chr.16: 57382755-57382755 on Build GRCh38
rs777497510 Chr.16: 57382752-57382752 on Build GRCh38
rs542418023 Chr.16: 57382698-57382698 on Build GRCh38
rs763320101 Chr.16: 57382686-57382686 on Build GRCh38
rs771178265 Chr.16: 57382659-57382659 on Build GRCh38
rs765773795 Chr.16: 57382675-57382675 on Build GRCh38
rs574947825 Chr.16: 57382694-57382694 on Build GRCh38
rs747983480 Chr.16: 57382640-57382640 on Build GRCh38
rs755935146 Chr.16: 57382647-57382647 on Build GRCh38
rs150690781 Chr.16: 57382575-57382575 on Build GRCh38
rs373175260 Chr.16: 57382574-57382574 on Build GRCh38
rs776630809 Chr.16: 57382660-57382660 on Build GRCh38
rs748044610 Chr.16: 57382743-57382743 on Build GRCh38
rs779020106 Chr.16: 57382722-57382722 on Build GRCh38
rs765718689 Chr.16: 57382602-57382602 on Build GRCh38
rs766308967 Chr.16: 57382587-57382587 on Build GRCh38
rs747130836 Chr.16: 57382657-57382657 on Build GRCh38
rs758722276 Chr.16: 57382612-57382612 on Build GRCh38
rs749987999 Chr.16: 57382622-57382622 on Build GRCh38
rs374046759 Chr.16: 57382623-57382623 on Build GRCh38
rs750170097 Chr.16: 57382639-57382639 on Build GRCh38
rs771108340 Chr.16: 57382757-57382757 on Build GRCh38
rs1010171372 Chr.16: 57382739-57382739 on Build GRCh38
rs559282444 Chr.16: 57382727-57382727 on Build GRCh38
rs371575579 Chr.16: 57382627-57382627 on Build GRCh38
rs758275490 Chr.16: 57382744-57382744 on Build GRCh38
rs749198932 Chr.16: 57382705-57382705 on Build GRCh38
rs541472573 Chr.16: 57382685-57382685 on Build GRCh38
rs149398209 Chr.16: 57382634-57382634 on Build GRCh38
rs12928112 Chr.16: 57382764-57382764 on Build GRCh38
rs376106665 Chr.16: 57382591-57382591 on Build GRCh38
rs776881670 Chr.16: 57382758-57382758 on Build GRCh38
rs777198174 Chr.16: 57382589-57382589 on Build GRCh38
rs755018361 Chr.16: 57382638-57382638 on Build GRCh38
rs772459306 Chr.16: 57382569-57382569 on Build GRCh38
rs138968060 Chr.16: 57382618-57382618 on Build GRCh38
rs779500193 Chr.16: 57382631-57382631 on Build GRCh38
rs759588929 Chr.16: 57382673-57382673 on Build GRCh38
rs149904106 Chr.16: 57382693-57382693 on Build GRCh38
rs558904875 Chr.16: 57382648-57382648 on Build GRCh38
rs753109663 Chr.16: 57382611-57382611 on Build GRCh38

View Full Product Details