Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158671597-158671840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf46
Entrez Gene ID
201725
Gene Name
chromosome 4 open reading frame 46
Gene Aliases
RCDG1
UniGene
Hs.380920
Gene Chromosome Location
Chr.4: 158666675-158672255 on Build GRCh38
Transcripts
NM_001008393.3 NR_077234.1 NR_077235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
106372 c.124C>T p.P42S Chr.4: 158671678-158671678 on Build GRCh38
5709137 c.50C>G p.P17R Chr.4: 158671752-158671752 on Build GRCh38
733291 c.77C>G p.S26C Chr.4: 158671725-158671725 on Build GRCh38
5978923 c.70G>T p.D24Y Chr.4: 158671732-158671732 on Build GRCh38
1428301 c.122T>C p.V41A Chr.4: 158671680-158671680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572351775 Chr.4: 158671622-158671622 on Build GRCh38
rs553203589 Chr.4: 158671649-158671649 on Build GRCh38
rs145167320 Chr.4: 158671723-158671723 on Build GRCh38
rs777098728 Chr.4: 158671692-158671692 on Build GRCh38
rs890415256 Chr.4: 158671750-158671750 on Build GRCh38
rs781514500 Chr.4: 158671760-158671760 on Build GRCh38
rs4596202 Chr.4: 158671796-158671796 on Build GRCh38
rs755273406 Chr.4: 158671740-158671740 on Build GRCh38
rs544594848 Chr.4: 158671799-158671799 on Build GRCh38
rs761401253 Chr.4: 158671821-158671821 on Build GRCh38
rs747346810 Chr.4: 158671730-158671730 on Build GRCh38
rs934321799 Chr.4: 158671629-158671629 on Build GRCh38
rs373406193 Chr.4: 158671670-158671670 on Build GRCh38
rs148787311 Chr.4: 158671709-158671709 on Build GRCh38
rs746373666 Chr.4: 158671638-158671638 on Build GRCh38
rs146620946 Chr.4: 158671691-158671691 on Build GRCh38
rs750487027 Chr.4: 158671636-158671636 on Build GRCh38
rs747196215 Chr.4: 158671651-158671651 on Build GRCh38
rs141418697 Chr.4: 158671702-158671702 on Build GRCh38
rs866237356 Chr.4: 158671683-158671683 on Build GRCh38
rs922235644 Chr.4: 158671671-158671671 on Build GRCh38
rs575518153 Chr.4: 158671797-158671797 on Build GRCh38
rs762346639 Chr.4: 158671720-158671731 on Build GRCh38
rs773359219 Chr.4: 158671786-158671786 on Build GRCh38
rs149785447 Chr.4: 158671666-158671666 on Build GRCh38
rs140720883 Chr.4: 158671639-158671639 on Build GRCh38
rs369968557 Chr.4: 158671656-158671656 on Build GRCh38
rs925623672 Chr.4: 158671615-158671615 on Build GRCh38
rs373903406 Chr.4: 158671811-158671811 on Build GRCh38
rs771997932 Chr.4: 158671793-158671793 on Build GRCh38
rs1038393624 Chr.4: 158671669-158671669 on Build GRCh38
rs867000557 Chr.4: 158671753-158671753 on Build GRCh38
rs758832408 Chr.4: 158671722-158671722 on Build GRCh38
rs140932563 Chr.4: 158671678-158671678 on Build GRCh38
rs776327084 Chr.4: 158671815-158671815 on Build GRCh38
rs750945797 Chr.4: 158671710-158671710 on Build GRCh38
rs142913539 Chr.4: 158671690-158671690 on Build GRCh38
rs760268980 Chr.4: 158671700-158671700 on Build GRCh38
rs376877943 Chr.4: 158671792-158671792 on Build GRCh38
rs776593810 Chr.4: 158671663-158671663 on Build GRCh38
rs993478281 Chr.4: 158671810-158671810 on Build GRCh38
rs989473296 Chr.4: 158671631-158671631 on Build GRCh38
rs148543446 Chr.4: 158671687-158671687 on Build GRCh38
rs748375809 Chr.4: 158671770-158671770 on Build GRCh38
rs753342205 Chr.4: 158671620-158671620 on Build GRCh38
rs926767530 Chr.4: 158671761-158671761 on Build GRCh38
rs770056558 Chr.4: 158671784-158671784 on Build GRCh38
rs1007763955 Chr.4: 158671820-158671820 on Build GRCh38
rs939393039 Chr.4: 158671766-158671766 on Build GRCh38
rs774495885 Chr.4: 158671667-158671667 on Build GRCh38
rs753459998 Chr.4: 158671704-158671704 on Build GRCh38
rs145770834 Chr.4: 158671668-158671668 on Build GRCh38

View Full Product Details