Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120497876-120498143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL1
Entrez Gene ID
8655
Gene Name
dynein light chain LC8-type 1
Gene Aliases
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1
UniGene
Hs.5120
Gene Chromosome Location
Chr.12: 120469857-120498495 on Build GRCh38
Transcripts
NM_001037494.1 NM_001037495.1 NM_003746.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1299004 c.180G>C p.R60S Chr.12: 120498120-120498120 on Build GRCh38
3811225 c.177G>A p.G59G Chr.12: 120498117-120498117 on Build GRCh38
6452065 c.187G>A p.G63S Chr.12: 120498127-120498127 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774659857 Chr.12: 120498036-120498036 on Build GRCh38
rs752949866 Chr.12: 120498047-120498047 on Build GRCh38
rs201501190 Chr.12: 120498048-120498048 on Build GRCh38
rs779642950 Chr.12: 120498075-120498075 on Build GRCh38
rs763431760 Chr.12: 120498025-120498025 on Build GRCh38
rs912244772 Chr.12: 120498112-120498112 on Build GRCh38
rs147364479 Chr.12: 120498003-120498003 on Build GRCh38
rs1012246368 Chr.12: 120497933-120497933 on Build GRCh38
rs768206330 Chr.12: 120498111-120498111 on Build GRCh38
rs375698180 Chr.12: 120498061-120498061 on Build GRCh38
rs188270446 Chr.12: 120498054-120498054 on Build GRCh38
rs759809842 Chr.12: 120498037-120498037 on Build GRCh38
rs970990010 Chr.12: 120497938-120497938 on Build GRCh38
rs552616110 Chr.12: 120497929-120497929 on Build GRCh38
rs750114378 Chr.12: 120498067-120498067 on Build GRCh38
rs370143651 Chr.12: 120498057-120498057 on Build GRCh38
rs573112768 Chr.12: 120497916-120497916 on Build GRCh38
rs764235939 Chr.12: 120498049-120498049 on Build GRCh38
rs771308218 Chr.12: 120498026-120498026 on Build GRCh38
rs1025429655 Chr.12: 120497934-120497934 on Build GRCh38
rs542247229 Chr.12: 120497975-120497975 on Build GRCh38
rs767577870 Chr.12: 120498046-120498046 on Build GRCh38
rs919639955 Chr.12: 120498056-120498056 on Build GRCh38
rs923696768 Chr.12: 120497998-120497998 on Build GRCh38
rs959590125 Chr.12: 120497906-120497906 on Build GRCh38
rs985136854 Chr.12: 120498098-120498098 on Build GRCh38
rs768834496 Chr.12: 120497974-120497974 on Build GRCh38
rs758080162 Chr.12: 120498070-120498070 on Build GRCh38
rs559799087 Chr.12: 120497910-120497910 on Build GRCh38
rs199508653 Chr.12: 120498071-120498071 on Build GRCh38
rs200418127 Chr.12: 120498063-120498063 on Build GRCh38
rs530739484 Chr.12: 120498020-120498020 on Build GRCh38
rs142458935 Chr.12: 120498105-120498105 on Build GRCh38
rs967699212 Chr.12: 120497945-120497945 on Build GRCh38

View Full Product Details