Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38937434-38937659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF33
Entrez Gene ID
100271715
Gene Name
Rho guanine nucleotide exchange factor 33
Gene Aliases
-
UniGene
Hs.99841
Gene Chromosome Location
Chr.2: 38919363-38975449 on Build GRCh38
Transcripts
NM_001145451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6393218 c.712C>A p.H238N Chr.2: 38937481-38937481 on Build GRCh38
6395347 c.765C>A p.N255K Chr.2: 38937534-38937534 on Build GRCh38
7410801 c.699C>T p.Y233Y Chr.2: 38937468-38937468 on Build GRCh38
3782198 c.700G>A p.V234I Chr.2: 38937469-38937469 on Build GRCh38
5655438 c.673G>C p.D225H Chr.2: 38937442-38937442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891432113 Chr.2: 38937469-38937469 on Build GRCh38
rs768029217 Chr.2: 38937575-38937575 on Build GRCh38
rs146076365 Chr.2: 38937535-38937535 on Build GRCh38
rs761473413 Chr.2: 38937525-38937525 on Build GRCh38
rs1011820519 Chr.2: 38937631-38937631 on Build GRCh38
rs976685412 Chr.2: 38937589-38937589 on Build GRCh38
rs768675278 Chr.2: 38937630-38937630 on Build GRCh38
rs950655945 Chr.2: 38937612-38937612 on Build GRCh38
rs548940033 Chr.2: 38937487-38937487 on Build GRCh38
rs567777256 Chr.2: 38937493-38937493 on Build GRCh38
rs200857822 Chr.2: 38937558-38937558 on Build GRCh38
rs1003052233 Chr.2: 38937583-38937583 on Build GRCh38
rs1047196206 Chr.2: 38937574-38937574 on Build GRCh38
rs776406798 Chr.2: 38937524-38937524 on Build GRCh38
rs745771806 Chr.2: 38937458-38937458 on Build GRCh38
rs1009962010 Chr.2: 38937475-38937475 on Build GRCh38
rs769393812 Chr.2: 38937534-38937534 on Build GRCh38
rs779748452 Chr.2: 38937484-38937484 on Build GRCh38
rs372530806 Chr.2: 38937468-38937468 on Build GRCh38
rs868183469 Chr.2: 38937462-38937462 on Build GRCh38
rs1030350957 Chr.2: 38937597-38937597 on Build GRCh38
rs1034892684 Chr.2: 38937611-38937611 on Build GRCh38
rs1007171726 Chr.2: 38937552-38937552 on Build GRCh38
rs894641362 Chr.2: 38937628-38937628 on Build GRCh38
rs983542640 Chr.2: 38937615-38937615 on Build GRCh38
rs753006512 Chr.2: 38937577-38937577 on Build GRCh38

View Full Product Details