Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23836946-23837205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL3
Entrez Gene ID
91319
Gene Name
derlin 3
Gene Aliases
C22orf14, IZP6, LLN2, derlin-3
UniGene
Hs.593679
Gene Chromosome Location
Chr.22: 23834503-23839344 on Build GRCh38
Transcripts
NM_001002862.2 NM_001135751.1 NM_198440.3 XM_011530505.2 XM_011530506.2 XM_017029078.1 XM_017029079.1 XM_017029080.1 XM_017029081.1 XM_017029082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1725617 c.568C>A p.P190T Chr.22: 23837110-23837110 on Build GRCh38
81085 c.561C>T p.D187D Chr.22: 23837117-23837117 on Build GRCh38
3405546 c.542T>C p.I181T Chr.22: 23837136-23837136 on Build GRCh38
1682146 c.591G>T p.R197S Chr.22: 23837087-23837087 on Build GRCh38
5548712 c.530C>T p.A177V Chr.22: 23837148-23837148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573264892 Chr.22: 23837037-23837037 on Build GRCh38
rs755702007 Chr.22: 23837124-23837124 on Build GRCh38
rs372554198 Chr.22: 23836999-23836999 on Build GRCh38
rs776575410 Chr.22: 23837087-23837087 on Build GRCh38
rs768605408 Chr.22: 23836993-23836993 on Build GRCh38
rs758785610 Chr.22: 23837117-23837117 on Build GRCh38
rs765430678 Chr.22: 23837011-23837011 on Build GRCh38
rs780721767 Chr.22: 23836978-23836978 on Build GRCh38
rs117846678 Chr.22: 23837147-23837147 on Build GRCh38
rs374017529 Chr.22: 23836986-23836986 on Build GRCh38
rs886377767 Chr.22: 23837164-23837164 on Build GRCh38
rs866535475 Chr.22: 23837184-23837184 on Build GRCh38
rs771983769 Chr.22: 23837078-23837078 on Build GRCh38
rs759983083 Chr.22: 23836974-23836990 on Build GRCh38
rs943107377 Chr.22: 23837079-23837079 on Build GRCh38
rs775902487 Chr.22: 23837109-23837109 on Build GRCh38
rs1047624710 Chr.22: 23837163-23837163 on Build GRCh38
rs35635247 Chr.22: 23837171-23837172 on Build GRCh38
rs779167583 Chr.22: 23837002-23837002 on Build GRCh38
rs746897426 Chr.22: 23837169-23837169 on Build GRCh38
rs748051720 Chr.22: 23836998-23836998 on Build GRCh38
rs778292185 Chr.22: 23837021-23837021 on Build GRCh38
rs1037449404 Chr.22: 23837084-23837084 on Build GRCh38
rs567035571 Chr.22: 23836984-23836984 on Build GRCh38
rs750541274 Chr.22: 23837004-23837005 on Build GRCh38
rs748920443 Chr.22: 23837059-23837059 on Build GRCh38
rs377205513 Chr.22: 23837010-23837010 on Build GRCh38
rs112734547 Chr.22: 23837156-23837156 on Build GRCh38
rs754607537 Chr.22: 23837141-23837141 on Build GRCh38
rs769929696 Chr.22: 23837104-23837104 on Build GRCh38
rs754811736 Chr.22: 23837065-23837065 on Build GRCh38
rs375769198 Chr.22: 23837076-23837076 on Build GRCh38
rs552271969 Chr.22: 23837035-23837035 on Build GRCh38
rs113967591 Chr.22: 23837157-23837157 on Build GRCh38
rs372138437 Chr.22: 23836974-23836974 on Build GRCh38
rs554531445 Chr.22: 23837139-23837139 on Build GRCh38
rs765741009 Chr.22: 23836985-23836994 on Build GRCh38
rs144605010 Chr.22: 23837170-23837170 on Build GRCh38
rs776754528 Chr.22: 23837006-23837006 on Build GRCh38
rs774324884 Chr.22: 23836995-23836995 on Build GRCh38
rs751848498 Chr.22: 23837174-23837175 on Build GRCh38
rs537893405 Chr.22: 23837120-23837120 on Build GRCh38
rs777838393 Chr.22: 23837165-23837166 on Build GRCh38
rs756875827 Chr.22: 23836975-23836975 on Build GRCh38
rs763235393 Chr.22: 23837111-23837111 on Build GRCh38
rs764620031 Chr.22: 23837117-23837117 on Build GRCh38
rs780276958 Chr.22: 23837185-23837185 on Build GRCh38
rs764530283 Chr.22: 23837029-23837029 on Build GRCh38
rs913509426 Chr.22: 23837055-23837055 on Build GRCh38
rs369004034 Chr.22: 23837116-23837116 on Build GRCh38
rs778382348 Chr.22: 23836968-23836968 on Build GRCh38
rs372803050 Chr.22: 23837050-23837050 on Build GRCh38
rs1051885475 Chr.22: 23837106-23837106 on Build GRCh38
rs1024274433 Chr.22: 23837180-23837180 on Build GRCh38
rs775746663 Chr.22: 23837012-23837012 on Build GRCh38
rs7290412 Chr.22: 23836979-23836979 on Build GRCh38
rs200374286 Chr.22: 23837048-23837048 on Build GRCh38
rs201370576 Chr.22: 23836989-23836989 on Build GRCh38
rs140089660 Chr.22: 23837151-23837151 on Build GRCh38
rs752419047 Chr.22: 23837148-23837148 on Build GRCh38
rs568972029 Chr.22: 23837042-23837042 on Build GRCh38
rs774798429 Chr.22: 23837119-23837119 on Build GRCh38
rs924853558 Chr.22: 23837044-23837044 on Build GRCh38
rs753023723 Chr.22: 23837083-23837083 on Build GRCh38
rs757825964 Chr.22: 23837030-23837030 on Build GRCh38
rs370046312 Chr.22: 23837020-23837020 on Build GRCh38
rs745917709 Chr.22: 23837099-23837099 on Build GRCh38
rs144407339 Chr.22: 23837123-23837123 on Build GRCh38

View Full Product Details