Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120196726-120196999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
UniGene
Hs.409662
Gene Chromosome Location
Chr.8: 120124468-120373575 on Build GRCh38
Transcripts
NM_021110.3 XM_005251059.3 XM_006716651.3 XM_017013809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3165623 c.490G>A p.D164N Chr.8: 120196844-120196844 on Build GRCh38
4760722 c.496T>G p.S166A Chr.8: 120196850-120196850 on Build GRCh38
3644661 c.483C>T p.I161I Chr.8: 120196837-120196837 on Build GRCh38
3644662 c.489C>T p.V163V Chr.8: 120196843-120196843 on Build GRCh38
3165625 c.540_541insT p.L181fs*23 Chr.8: 120196894-120196895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747499046 Chr.8: 120196777-120196777 on Build GRCh38
rs143945195 Chr.8: 120196853-120196853 on Build GRCh38
rs746732484 Chr.8: 120196814-120196814 on Build GRCh38
rs267601747 Chr.8: 120196894-120196894 on Build GRCh38
rs749490638 Chr.8: 120196852-120196852 on Build GRCh38
rs544217331 Chr.8: 120196821-120196821 on Build GRCh38
rs796912542 Chr.8: 120196793-120196793 on Build GRCh38
rs986793698 Chr.8: 120196846-120196846 on Build GRCh38
rs912674451 Chr.8: 120196867-120196867 on Build GRCh38
rs376105584 Chr.8: 120196797-120196797 on Build GRCh38
rs756025821 Chr.8: 120196945-120196945 on Build GRCh38
rs762164576 Chr.8: 120196879-120196879 on Build GRCh38
rs764685863 Chr.8: 120196890-120196890 on Build GRCh38
rs113064195 Chr.8: 120196767-120196767 on Build GRCh38
rs750355331 Chr.8: 120196941-120196941 on Build GRCh38
rs762324152 Chr.8: 120196764-120196764 on Build GRCh38
rs61729463 Chr.8: 120196802-120196802 on Build GRCh38
rs777735334 Chr.8: 120196808-120196808 on Build GRCh38
rs149492812 Chr.8: 120196843-120196843 on Build GRCh38
rs958214735 Chr.8: 120196820-120196820 on Build GRCh38
rs924461333 Chr.8: 120196972-120196972 on Build GRCh38
rs768183755 Chr.8: 120196918-120196918 on Build GRCh38
rs767948764 Chr.8: 120196766-120196766 on Build GRCh38
rs752203859 Chr.8: 120196909-120196909 on Build GRCh38
rs372816216 Chr.8: 120196773-120196773 on Build GRCh38
rs749325378 Chr.8: 120196958-120196958 on Build GRCh38
rs946062646 Chr.8: 120196845-120196845 on Build GRCh38
rs764394379 Chr.8: 120196759-120196759 on Build GRCh38
rs774704813 Chr.8: 120196877-120196877 on Build GRCh38
rs768073565 Chr.8: 120196882-120196882 on Build GRCh38
rs780009646 Chr.8: 120196954-120196954 on Build GRCh38
rs931566070 Chr.8: 120196922-120196922 on Build GRCh38
rs746008979 Chr.8: 120196838-120196838 on Build GRCh38
rs193209164 Chr.8: 120196887-120196887 on Build GRCh38
rs748565100 Chr.8: 120196976-120196976 on Build GRCh38
rs115892986 Chr.8: 120196920-120196920 on Build GRCh38
rs779098096 Chr.8: 120196970-120196970 on Build GRCh38
rs775596675 Chr.8: 120196848-120196848 on Build GRCh38
rs556415250 Chr.8: 120196837-120196837 on Build GRCh38
rs1047344104 Chr.8: 120196940-120196940 on Build GRCh38
rs759127092 Chr.8: 120196888-120196888 on Build GRCh38
rs763832906 Chr.8: 120196919-120196919 on Build GRCh38
rs188819739 Chr.8: 120196780-120196780 on Build GRCh38
rs762528880 Chr.8: 120196911-120196911 on Build GRCh38
rs2305597 Chr.8: 120196761-120196761 on Build GRCh38
rs781319319 Chr.8: 120196939-120196939 on Build GRCh38
rs757095908 Chr.8: 120196936-120196936 on Build GRCh38
rs1013198893 Chr.8: 120196760-120196760 on Build GRCh38
rs768936494 Chr.8: 120196965-120196965 on Build GRCh38
rs768805792 Chr.8: 120196868-120196868 on Build GRCh38

View Full Product Details