Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159713953-159714178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRP
Entrez Gene ID
1401
Gene Name
C-reactive protein, pentraxin-related
Gene Aliases
PTX1
UniGene
Hs.709456
Gene Chromosome Location
Chr.1: 159712289-159714623 on Build GRCh38
Transcripts
NM_000567.2 XM_011509207.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844136 c.197_198GG>AA p.G66E Chr.1: 159714002-159714003 on Build GRCh38
5844135 c.203G>A p.S68N Chr.1: 159713997-159713997 on Build GRCh38
3802429 c.185C>G p.S62C Chr.1: 159714015-159714015 on Build GRCh38
5911281 c.86T>A p.F29Y Chr.1: 159714114-159714114 on Build GRCh38
7075937 c.246C>G p.L82L Chr.1: 159713954-159713954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769624182 Chr.1: 159714139-159714139 on Build GRCh38
rs201563359 Chr.1: 159714051-159714051 on Build GRCh38
rs922194613 Chr.1: 159714090-159714090 on Build GRCh38
rs36061058 Chr.1: 159714071-159714071 on Build GRCh38
rs373675212 Chr.1: 159714104-159714104 on Build GRCh38
rs760410118 Chr.1: 159713988-159713988 on Build GRCh38
rs766040036 Chr.1: 159714017-159714017 on Build GRCh38
rs745977455 Chr.1: 159714043-159714043 on Build GRCh38
rs116776267 Chr.1: 159714066-159714066 on Build GRCh38
rs775554201 Chr.1: 159714054-159714054 on Build GRCh38
rs776402087 Chr.1: 159714006-159714006 on Build GRCh38
rs780247434 Chr.1: 159714118-159714118 on Build GRCh38
rs35500644 Chr.1: 159714095-159714095 on Build GRCh38
rs780808995 Chr.1: 159714131-159714131 on Build GRCh38
rs370716776 Chr.1: 159714086-159714086 on Build GRCh38
rs77832441 Chr.1: 159714024-159714024 on Build GRCh38
rs775316926 Chr.1: 159713987-159713987 on Build GRCh38
rs749938616 Chr.1: 159714011-159714011 on Build GRCh38
rs567820071 Chr.1: 159714082-159714082 on Build GRCh38
rs570340515 Chr.1: 159714061-159714061 on Build GRCh38
rs766797643 Chr.1: 159714152-159714155 on Build GRCh38
rs769752475 Chr.1: 159714072-159714072 on Build GRCh38
rs751271674 Chr.1: 159714023-159714023 on Build GRCh38
rs757801362 Chr.1: 159714096-159714096 on Build GRCh38
rs34672691 Chr.1: 159714001-159714001 on Build GRCh38
rs779071283 Chr.1: 159714037-159714037 on Build GRCh38
rs867291501 Chr.1: 159714084-159714084 on Build GRCh38
rs553010709 Chr.1: 159714141-159714141 on Build GRCh38
rs747295746 Chr.1: 159714055-159714055 on Build GRCh38
rs768322386 Chr.1: 159714065-159714065 on Build GRCh38
rs779587324 Chr.1: 159714102-159714102 on Build GRCh38
rs759484412 Chr.1: 159714124-159714124 on Build GRCh38
rs749749899 Chr.1: 159714025-159714025 on Build GRCh38
rs143101792 Chr.1: 159714026-159714026 on Build GRCh38
rs377351051 Chr.1: 159714007-159714007 on Build GRCh38
rs747142678 Chr.1: 159714124-159714124 on Build GRCh38
rs759295403 Chr.1: 159714085-159714085 on Build GRCh38
rs762941084 Chr.1: 159714143-159714143 on Build GRCh38
rs373463561 Chr.1: 159713997-159713997 on Build GRCh38
rs762424423 Chr.1: 159714073-159714073 on Build GRCh38
rs765949437 Chr.1: 159714075-159714075 on Build GRCh38
rs768926770 Chr.1: 159714128-159714128 on Build GRCh38
rs148534477 Chr.1: 159714106-159714106 on Build GRCh38
rs1026679333 Chr.1: 159714110-159714110 on Build GRCh38
rs145295141 Chr.1: 159714012-159714012 on Build GRCh38
rs764668650 Chr.1: 159714008-159714008 on Build GRCh38

View Full Product Details