Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42104477-42104726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf132
Entrez Gene ID
647024
Gene Name
chromosome 6 open reading frame 132
Gene Aliases
bA7K24.2
UniGene
Hs.444277
Gene Chromosome Location
Chr.6: 42101119-42142977 on Build GRCh38
Transcripts
NM_001164446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6859281 c.3307C>T p.R1103C Chr.6: 42104605-42104605 on Build GRCh38
5474940 c.3205G>A p.G1069R Chr.6: 42104707-42104707 on Build GRCh38
6193289 c.3288C>T p.P1096P Chr.6: 42104624-42104624 on Build GRCh38
1742888 c.3247G>A p.G1083S Chr.6: 42104665-42104665 on Build GRCh38
7275197 c.3249C>T p.G1083G Chr.6: 42104663-42104663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048042861 Chr.6: 42104533-42104533 on Build GRCh38
rs758536814 Chr.6: 42104701-42104701 on Build GRCh38
rs949273220 Chr.6: 42104532-42104532 on Build GRCh38
rs1010374197 Chr.6: 42104642-42104642 on Build GRCh38
rs1010140562 Chr.6: 42104643-42104643 on Build GRCh38
rs975465598 Chr.6: 42104575-42104575 on Build GRCh38
rs764883179 Chr.6: 42104569-42104616 on Build GRCh38
rs907884819 Chr.6: 42104591-42104591 on Build GRCh38
rs921233239 Chr.6: 42104588-42104588 on Build GRCh38
rs567997884 Chr.6: 42104651-42104651 on Build GRCh38
rs779454967 Chr.6: 42104658-42104658 on Build GRCh38
rs983889500 Chr.6: 42104578-42104578 on Build GRCh38
rs1037325383 Chr.6: 42104562-42104562 on Build GRCh38
rs867094463 Chr.6: 42104592-42104592 on Build GRCh38
rs536844087 Chr.6: 42104536-42104536 on Build GRCh38
rs1052044789 Chr.6: 42104624-42104624 on Build GRCh38
rs1054212635 Chr.6: 42104629-42104629 on Build GRCh38
rs921984969 Chr.6: 42104618-42104618 on Build GRCh38
rs1040437107 Chr.6: 42104612-42104612 on Build GRCh38
rs1041995526 Chr.6: 42104683-42104683 on Build GRCh38
rs968791132 Chr.6: 42104652-42104652 on Build GRCh38
rs753096473 Chr.6: 42104686-42104686 on Build GRCh38
rs866592048 Chr.6: 42104521-42104521 on Build GRCh38
rs9471761 Chr.6: 42104641-42104641 on Build GRCh38
rs1002919410 Chr.6: 42104700-42104700 on Build GRCh38
rs527914512 Chr.6: 42104565-42104565 on Build GRCh38
rs990978176 Chr.6: 42104505-42104505 on Build GRCh38
rs959537627 Chr.6: 42104498-42104498 on Build GRCh38
rs755583581 Chr.6: 42104654-42104654 on Build GRCh38
rs931848033 Chr.6: 42104619-42104619 on Build GRCh38
rs920886624 Chr.6: 42104511-42104511 on Build GRCh38
rs1000322711 Chr.6: 42104675-42104675 on Build GRCh38
rs906884894 Chr.6: 42104692-42104692 on Build GRCh38
rs944681840 Chr.6: 42104602-42104602 on Build GRCh38

View Full Product Details