Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55678355-55678594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899962 c.290C>T p.P97L Chr.19: 55678584-55678584 on Build GRCh38
4608794 c.245G>T p.C82F Chr.19: 55678539-55678539 on Build GRCh38
7351735 c.233-1G>C p.? Chr.19: 55678526-55678526 on Build GRCh38
6274791 c.267G>T p.S89S Chr.19: 55678561-55678561 on Build GRCh38
2969204 c.284T>C p.I95T Chr.19: 55678578-55678578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776270294 Chr.19: 55678494-55678494 on Build GRCh38
rs141127387 Chr.19: 55678552-55678552 on Build GRCh38
rs770936438 Chr.19: 55678474-55678474 on Build GRCh38
rs543718478 Chr.19: 55678484-55678484 on Build GRCh38
rs1016485647 Chr.19: 55678487-55678487 on Build GRCh38
rs939772775 Chr.19: 55678507-55678507 on Build GRCh38
rs1000880044 Chr.19: 55678376-55678376 on Build GRCh38
rs1014121359 Chr.19: 55678456-55678456 on Build GRCh38
rs754039807 Chr.19: 55678539-55678539 on Build GRCh38
rs774429557 Chr.19: 55678478-55678478 on Build GRCh38
rs969553763 Chr.19: 55678472-55678472 on Build GRCh38
rs542251105 Chr.19: 55678561-55678561 on Build GRCh38
rs757113261 Chr.19: 55678571-55678571 on Build GRCh38
rs769654129 Chr.19: 55678576-55678576 on Build GRCh38
rs762175445 Chr.19: 55678528-55678528 on Build GRCh38
rs1031380907 Chr.19: 55678380-55678380 on Build GRCh38
rs745634936 Chr.19: 55678573-55678573 on Build GRCh38
rs556954533 Chr.19: 55678537-55678537 on Build GRCh38
rs949500865 Chr.19: 55678530-55678530 on Build GRCh38
rs892463087 Chr.19: 55678439-55678439 on Build GRCh38
rs866543589 Chr.19: 55678416-55678416 on Build GRCh38
rs764202866 Chr.19: 55678522-55678522 on Build GRCh38
rs750752386 Chr.19: 55678551-55678551 on Build GRCh38
rs1025002366 Chr.19: 55678469-55678469 on Build GRCh38
rs781106190 Chr.19: 55678572-55678572 on Build GRCh38
rs759844592 Chr.19: 55678499-55678499 on Build GRCh38
rs778554427 Chr.19: 55678554-55678554 on Build GRCh38
rs965145321 Chr.19: 55678426-55678426 on Build GRCh38
rs73613271 Chr.19: 55678501-55678501 on Build GRCh38
rs554434480 Chr.19: 55678383-55678383 on Build GRCh38
rs540110122 Chr.19: 55678517-55678517 on Build GRCh38
rs373209112 Chr.19: 55678564-55678564 on Build GRCh38
rs201101921 Chr.19: 55678565-55678565 on Build GRCh38
rs963932233 Chr.19: 55678491-55678491 on Build GRCh38
rs184026465 Chr.19: 55678523-55678523 on Build GRCh38
rs768130917 Chr.19: 55678560-55678560 on Build GRCh38
rs752139537 Chr.19: 55678524-55678524 on Build GRCh38
rs976657089 Chr.19: 55678443-55678443 on Build GRCh38
rs918959681 Chr.19: 55678504-55678504 on Build GRCh38

View Full Product Details