Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122103427-122103688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254602 c.134C>T p.S45F Chr.3: 122103581-122103581 on Build GRCh38
6623928 c.217G>A p.E73K Chr.3: 122103664-122103664 on Build GRCh38
3845977 c.109G>A p.D37N Chr.3: 122103556-122103556 on Build GRCh38
3121182 c.235C>T p.H79Y Chr.3: 122103682-122103682 on Build GRCh38
6269189 c.212G>T p.G71V Chr.3: 122103659-122103659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570961036 Chr.3: 122103507-122103507 on Build GRCh38
rs773800947 Chr.3: 122103520-122103520 on Build GRCh38
rs747238810 Chr.3: 122103553-122103553 on Build GRCh38
rs1042991536 Chr.3: 122103482-122103482 on Build GRCh38
rs759696095 Chr.3: 122103651-122103651 on Build GRCh38
rs752867412 Chr.3: 122103503-122103504 on Build GRCh38
rs1005895615 Chr.3: 122103598-122103598 on Build GRCh38
rs559114894 Chr.3: 122103615-122103615 on Build GRCh38
rs947281461 Chr.3: 122103457-122103457 on Build GRCh38
rs780604915 Chr.3: 122103470-122103470 on Build GRCh38
rs866110754 Chr.3: 122103578-122103578 on Build GRCh38
rs766139636 Chr.3: 122103500-122103502 on Build GRCh38
rs1015889379 Chr.3: 122103606-122103606 on Build GRCh38
rs938518326 Chr.3: 122103534-122103534 on Build GRCh38
rs778676985 Chr.3: 122103545-122103545 on Build GRCh38
rs755571029 Chr.3: 122103509-122103509 on Build GRCh38
rs948647409 Chr.3: 122103475-122103475 on Build GRCh38
rs865917709 Chr.3: 122103617-122103617 on Build GRCh38
rs1047493821 Chr.3: 122103559-122103559 on Build GRCh38
rs763378254 Chr.3: 122103570-122103570 on Build GRCh38
rs376775826 Chr.3: 122103525-122103525 on Build GRCh38
rs931781143 Chr.3: 122103577-122103577 on Build GRCh38
rs149667487 Chr.3: 122103518-122103518 on Build GRCh38
rs561788422 Chr.3: 122103488-122103488 on Build GRCh38
rs539519506 Chr.3: 122103530-122103530 on Build GRCh38
rs1044318780 Chr.3: 122103505-122103505 on Build GRCh38
rs112508906 Chr.3: 122103555-122103555 on Build GRCh38
rs553416970 Chr.3: 122103579-122103579 on Build GRCh38

View Full Product Details