Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 74538058-74538291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L3B
Entrez Gene ID
552889
Gene Name
ataxin 7 like 3B
Gene Aliases
lnc-SCA7
UniGene
Hs.744849
Gene Chromosome Location
Chr.12: 74537771-74541452 on Build GRCh38
Transcripts
NM_001136262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4974024 c.4G>C p.E2Q Chr.12: 74538116-74538116 on Build GRCh38
7337563 c.156G>C p.E52D Chr.12: 74538268-74538268 on Build GRCh38
943403 c.81G>A p.E27E Chr.12: 74538193-74538193 on Build GRCh38
4211208 c.70G>T p.D24Y Chr.12: 74538182-74538182 on Build GRCh38
5640171 c.143delT p.L48fs*9 Chr.12: 74538255-74538255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760247730 Chr.12: 74538254-74538254 on Build GRCh38
rs1014461040 Chr.12: 74538268-74538268 on Build GRCh38
rs368832656 Chr.12: 74538260-74538260 on Build GRCh38
rs779423791 Chr.12: 74538143-74538143 on Build GRCh38
rs547208470 Chr.12: 74538085-74538085 on Build GRCh38
rs376619943 Chr.12: 74538229-74538229 on Build GRCh38
rs772016191 Chr.12: 74538244-74538244 on Build GRCh38
rs763709866 Chr.12: 74538258-74538258 on Build GRCh38
rs539551666 Chr.12: 74538099-74538099 on Build GRCh38
rs757635962 Chr.12: 74538137-74538137 on Build GRCh38
rs1008724897 Chr.12: 74538082-74538082 on Build GRCh38
rs753600487 Chr.12: 74538130-74538130 on Build GRCh38
rs758961994 Chr.12: 74538147-74538147 on Build GRCh38
rs747077103 Chr.12: 74538152-74538152 on Build GRCh38
rs781336704 Chr.12: 74538167-74538167 on Build GRCh38
rs775358181 Chr.12: 74538252-74538252 on Build GRCh38
rs763836975 Chr.12: 74538123-74538123 on Build GRCh38
rs556666725 Chr.12: 74538232-74538232 on Build GRCh38
rs570274148 Chr.12: 74538087-74538087 on Build GRCh38
rs372445812 Chr.12: 74538226-74538226 on Build GRCh38
rs776687077 Chr.12: 74538264-74538264 on Build GRCh38
rs760144320 Chr.12: 74538107-74538107 on Build GRCh38
rs768787523 Chr.12: 74538166-74538166 on Build GRCh38
rs774042592 Chr.12: 74538269-74538269 on Build GRCh38
rs750854810 Chr.12: 74538145-74538145 on Build GRCh38
rs759298204 Chr.12: 74538237-74538237 on Build GRCh38
rs768585533 Chr.12: 74538091-74538091 on Build GRCh38
rs770729351 Chr.12: 74538079-74538079 on Build GRCh38
rs770676411 Chr.12: 74538230-74538230 on Build GRCh38
rs780471674 Chr.12: 74538148-74538148 on Build GRCh38
rs970963646 Chr.12: 74538206-74538206 on Build GRCh38

View Full Product Details