Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122216287-122216560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
UniGene
Hs.169611
Gene Chromosome Location
Chr.12: 122207662-122227534 on Build GRCh38
Transcripts
NM_001278302.1 NM_001278303.1 NM_001278304.1 NM_001278342.1 NM_019887.5 NM_138930.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5704321 c.523G>C p.G175R Chr.12: 122216488-122216488 on Build GRCh38
7231697 c.523+8C>G p.? Chr.12: 122216480-122216480 on Build GRCh38
5106992 c.502G>A p.A168T Chr.12: 122216509-122216509 on Build GRCh38
1322691 c.459G>A p.L153L Chr.12: 122216552-122216552 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3215200 Chr.12: 122216464-122216464 on Build GRCh38
rs190518526 Chr.12: 122216455-122216455 on Build GRCh38
rs994617815 Chr.12: 122216537-122216537 on Build GRCh38
rs761851919 Chr.12: 122216465-122216465 on Build GRCh38
rs775356846 Chr.12: 122216444-122216444 on Build GRCh38
rs754184666 Chr.12: 122216454-122216458 on Build GRCh38
rs79009021 Chr.12: 122216453-122216453 on Build GRCh38
rs1031837096 Chr.12: 122216401-122216401 on Build GRCh38
rs748596027 Chr.12: 122216518-122216518 on Build GRCh38
rs369663349 Chr.12: 122216464-122216464 on Build GRCh38
rs750574662 Chr.12: 122216470-122216470 on Build GRCh38
rs747231415 Chr.12: 122216337-122216337 on Build GRCh38
rs752215724 Chr.12: 122216446-122216447 on Build GRCh38
rs111589443 Chr.12: 122216454-122216454 on Build GRCh38
rs759028296 Chr.12: 122216455-122216455 on Build GRCh38
rs758998555 Chr.12: 122216522-122216522 on Build GRCh38
rs768900266 Chr.12: 122216455-122216458 on Build GRCh38
rs745376603 Chr.12: 122216535-122216535 on Build GRCh38
rs767491775 Chr.12: 122216469-122216469 on Build GRCh38
rs756276240 Chr.12: 122216484-122216484 on Build GRCh38
rs1056690931 Chr.12: 122216493-122216493 on Build GRCh38
rs977305542 Chr.12: 122216330-122216330 on Build GRCh38
rs770044737 Chr.12: 122216454-122216456 on Build GRCh38
rs576580943 Chr.12: 122216453-122216454 on Build GRCh38
rs776267494 Chr.12: 122216438-122216438 on Build GRCh38
rs747098107 Chr.12: 122216455-122216459 on Build GRCh38
rs776739128 Chr.12: 122216516-122216516 on Build GRCh38
rs755360766 Chr.12: 122216501-122216501 on Build GRCh38
rs112300336 Chr.12: 122216486-122216486 on Build GRCh38
rs199768401 Chr.12: 122216539-122216539 on Build GRCh38
rs76932423 Chr.12: 122216338-122216338 on Build GRCh38
rs745489788 Chr.12: 122216441-122216441 on Build GRCh38
rs747606181 Chr.12: 122216454-122216455 on Build GRCh38
rs964156140 Chr.12: 122216323-122216323 on Build GRCh38
rs781399473 Chr.12: 122216421-122216421 on Build GRCh38
rs754165375 Chr.12: 122216498-122216498 on Build GRCh38
rs762870855 Chr.12: 122216452-122216452 on Build GRCh38
rs935073619 Chr.12: 122216371-122216371 on Build GRCh38
rs779372275 Chr.12: 122216508-122216508 on Build GRCh38
rs780273024 Chr.12: 122216455-122216457 on Build GRCh38
rs780506459 Chr.12: 122216529-122216529 on Build GRCh38
rs796100906 Chr.12: 122216454-122216455 on Build GRCh38
rs2271411 Chr.12: 122216454-122216454 on Build GRCh38
rs769584215 Chr.12: 122216443-122216443 on Build GRCh38
rs566598819 Chr.12: 122216395-122216395 on Build GRCh38
rs757597153 Chr.12: 122216454-122216457 on Build GRCh38
rs766596915 Chr.12: 122216490-122216490 on Build GRCh38
rs780257614 Chr.12: 122216317-122216317 on Build GRCh38
rs770313143 Chr.12: 122216437-122216437 on Build GRCh38

View Full Product Details