Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155763740-155763989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf33
Entrez Gene ID
285315
Gene Name
chromosome 3 open reading frame 33
Gene Aliases
AC3-33
UniGene
Hs.350846
Gene Chromosome Location
Chr.3: 155762612-155806287 on Build GRCh38
Transcripts
NM_001308229.1 NM_173657.2 XM_005247379.4 XM_011512710.2 XM_011512711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617109 c.358C>A p.L120M Chr.3: 155763897-155763897 on Build GRCh38
1632927 c.493G>A p.G165R Chr.3: 155763762-155763762 on Build GRCh38
3660290 c.505C>A p.L169M Chr.3: 155763897-155763897 on Build GRCh38
7230755 c.655G>A p.D219N Chr.3: 155763747-155763747 on Build GRCh38
7225589 c.582C>T p.Y194Y Chr.3: 155763820-155763820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773359213 Chr.3: 155763926-155763926 on Build GRCh38
rs780436730 Chr.3: 155763789-155763789 on Build GRCh38
rs771659485 Chr.3: 155763855-155763855 on Build GRCh38
rs7653384 Chr.3: 155763820-155763820 on Build GRCh38
rs766362355 Chr.3: 155763917-155763917 on Build GRCh38
rs755894625 Chr.3: 155763819-155763819 on Build GRCh38
rs745444960 Chr.3: 155763845-155763845 on Build GRCh38
rs758679808 Chr.3: 155763775-155763775 on Build GRCh38
rs575233706 Chr.3: 155763782-155763782 on Build GRCh38
rs115731681 Chr.3: 155763904-155763904 on Build GRCh38
rs778772192 Chr.3: 155763781-155763781 on Build GRCh38
rs1024755082 Chr.3: 155763772-155763772 on Build GRCh38
rs749161316 Chr.3: 155763830-155763830 on Build GRCh38
rs772069197 Chr.3: 155763858-155763858 on Build GRCh38
rs763138200 Chr.3: 155763947-155763947 on Build GRCh38
rs752474590 Chr.3: 155763792-155763792 on Build GRCh38
rs926529641 Chr.3: 155763882-155763882 on Build GRCh38
rs374247931 Chr.3: 155763915-155763915 on Build GRCh38
rs147770210 Chr.3: 155763903-155763903 on Build GRCh38
rs267599663 Chr.3: 155763895-155763895 on Build GRCh38
rs751458858 Chr.3: 155763931-155763933 on Build GRCh38
rs747337825 Chr.3: 155763879-155763879 on Build GRCh38
rs775150071 Chr.3: 155763870-155763870 on Build GRCh38
rs952664076 Chr.3: 155763773-155763773 on Build GRCh38
rs778285413 Chr.3: 155763843-155763843 on Build GRCh38
rs762988845 Chr.3: 155763769-155763769 on Build GRCh38
rs750597545 Chr.3: 155763774-155763774 on Build GRCh38
rs984130638 Chr.3: 155763796-155763796 on Build GRCh38
rs770945064 Chr.3: 155763836-155763836 on Build GRCh38

View Full Product Details