Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15537610-15537869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
UniGene
Hs.156452
Gene Chromosome Location
Chr.19: 15508487-15552317 on Build GRCh38
Transcripts
NM_173483.3 XM_011527692.2 XM_011527693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3529718 c.502C>T p.L168L Chr.19: 15537615-15537615 on Build GRCh38
3283844 c.534C>T p.S178S Chr.19: 15537647-15537647 on Build GRCh38
5520503 c.508C>T p.P170S Chr.19: 15537621-15537621 on Build GRCh38
3283845 c.538G>A p.D180N Chr.19: 15537651-15537651 on Build GRCh38
4075064 c.519G>T p.K173N Chr.19: 15537632-15537632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143598221 Chr.19: 15537799-15537799 on Build GRCh38
rs774213936 Chr.19: 15537677-15537677 on Build GRCh38
rs201743364 Chr.19: 15537798-15537798 on Build GRCh38
rs752264797 Chr.19: 15537829-15537829 on Build GRCh38
rs761697748 Chr.19: 15537691-15537691 on Build GRCh38
rs573750979 Chr.19: 15537785-15537785 on Build GRCh38
rs768585238 Chr.19: 15537675-15537675 on Build GRCh38
rs140657557 Chr.19: 15537840-15537840 on Build GRCh38
rs1005088726 Chr.19: 15537713-15537713 on Build GRCh38
rs576705212 Chr.19: 15537814-15537814 on Build GRCh38
rs16980531 Chr.19: 15537645-15537645 on Build GRCh38
rs982261190 Chr.19: 15537818-15537818 on Build GRCh38
rs758816247 Chr.19: 15537648-15537648 on Build GRCh38
rs571757470 Chr.19: 15537656-15537656 on Build GRCh38
rs780816103 Chr.19: 15537649-15537649 on Build GRCh38
rs534242787 Chr.19: 15537704-15537704 on Build GRCh38
rs370400762 Chr.19: 15537670-15537670 on Build GRCh38
rs764977269 Chr.19: 15537805-15537805 on Build GRCh38
rs759543514 Chr.19: 15537845-15537845 on Build GRCh38
rs772035186 Chr.19: 15537694-15537694 on Build GRCh38
rs766540379 Chr.19: 15537826-15537826 on Build GRCh38
rs754310081 Chr.19: 15537736-15537736 on Build GRCh38
rs556566002 Chr.19: 15537807-15537807 on Build GRCh38
rs758503533 Chr.19: 15537687-15537688 on Build GRCh38
rs935078250 Chr.19: 15537834-15537834 on Build GRCh38
rs372867266 Chr.19: 15537709-15537709 on Build GRCh38
rs192304348 Chr.19: 15537757-15537757 on Build GRCh38
rs148063219 Chr.19: 15537804-15537804 on Build GRCh38
rs148454445 Chr.19: 15537652-15537652 on Build GRCh38
rs1055995688 Chr.19: 15537811-15537811 on Build GRCh38
rs946071699 Chr.19: 15537761-15537761 on Build GRCh38
rs377373101 Chr.19: 15537647-15537647 on Build GRCh38

View Full Product Details