Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27216575-27216846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRAID
Entrez Gene ID
51374
Gene Name
all-trans retinoic acid induced differentiation factor
Gene Aliases
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18
UniGene
Hs.9527
Gene Chromosome Location
Chr.2: 27212031-27217178 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667594 c.750+1G>C p.? Chr.2: 27216621-27216621 on Build GRCh38
5058849 c.706T>C p.C236R Chr.2: 27216576-27216576 on Build GRCh38
1019544 c.746G>A p.R249H Chr.2: 27216616-27216616 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778773190 Chr.2: 27216612-27216612 on Build GRCh38
rs781457675 Chr.2: 27216646-27216646 on Build GRCh38
rs757188443 Chr.2: 27216608-27216608 on Build GRCh38
rs755647683 Chr.2: 27216597-27216597 on Build GRCh38
rs546627525 Chr.2: 27216671-27216673 on Build GRCh38
rs771382401 Chr.2: 27216821-27216822 on Build GRCh38
rs541343233 Chr.2: 27216789-27216789 on Build GRCh38
rs41288811 Chr.2: 27216650-27216650 on Build GRCh38
rs563420711 Chr.2: 27216774-27216774 on Build GRCh38
rs775342548 Chr.2: 27216819-27216819 on Build GRCh38
rs112885738 Chr.2: 27216705-27216705 on Build GRCh38
rs745675289 Chr.2: 27216615-27216615 on Build GRCh38
rs769811139 Chr.2: 27216653-27216653 on Build GRCh38
rs767537168 Chr.2: 27216781-27216781 on Build GRCh38
rs559252443 Chr.2: 27216810-27216810 on Build GRCh38
rs748886262 Chr.2: 27216819-27216820 on Build GRCh38
rs771786328 Chr.2: 27216636-27216636 on Build GRCh38
rs184803940 Chr.2: 27216601-27216601 on Build GRCh38
rs778530773 Chr.2: 27216756-27216756 on Build GRCh38
rs999444721 Chr.2: 27216720-27216720 on Build GRCh38
rs139118951 Chr.2: 27216710-27216710 on Build GRCh38
rs375386400 Chr.2: 27216616-27216616 on Build GRCh38
rs138274094 Chr.2: 27216603-27216603 on Build GRCh38
rs140284109 Chr.2: 27216599-27216599 on Build GRCh38
rs767992196 Chr.2: 27216814-27216814 on Build GRCh38
rs201214418 Chr.2: 27216662-27216662 on Build GRCh38
rs893801073 Chr.2: 27216686-27216686 on Build GRCh38
rs771495187 Chr.2: 27216804-27216804 on Build GRCh38
rs763366667 Chr.2: 27216733-27216734 on Build GRCh38
rs773307733 Chr.2: 27216794-27216794 on Build GRCh38
rs751823502 Chr.2: 27216635-27216635 on Build GRCh38
rs769356644 Chr.2: 27216611-27216612 on Build GRCh38
rs774945277 Chr.2: 27216807-27216807 on Build GRCh38
rs762897150 Chr.2: 27216797-27216797 on Build GRCh38
rs372453045 Chr.2: 27216813-27216813 on Build GRCh38

View Full Product Details