Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9440440-9440713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
UniGene
Hs.515972
Gene Chromosome Location
Chr.2: 9423537-9473110 on Build GRCh38
Transcripts
NM_001321833.1 NM_001321834.1 NM_001321835.1 NM_001321836.1 NM_016207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4558364 c.763G>C p.E255Q Chr.2: 9440493-9440493 on Build GRCh38
1495001 c.833G>A p.C278Y Chr.2: 9440563-9440563 on Build GRCh38
1023679 c.862G>A p.A288T Chr.2: 9440592-9440592 on Build GRCh38
1023678 c.859A>C p.N287H Chr.2: 9440589-9440589 on Build GRCh38
6427596 c.925A>G p.S309G Chr.2: 9440655-9440655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035362399 Chr.2: 9440610-9440610 on Build GRCh38
rs763125673 Chr.2: 9440624-9440624 on Build GRCh38
rs768942697 Chr.2: 9440477-9440477 on Build GRCh38
rs140005782 Chr.2: 9440569-9440569 on Build GRCh38
rs140835115 Chr.2: 9440640-9440640 on Build GRCh38
rs780793543 Chr.2: 9440482-9440483 on Build GRCh38
rs780524166 Chr.2: 9440469-9440469 on Build GRCh38
rs144181988 Chr.2: 9440654-9440654 on Build GRCh38
rs755257190 Chr.2: 9440651-9440651 on Build GRCh38
rs1001396834 Chr.2: 9440546-9440546 on Build GRCh38
rs763897913 Chr.2: 9440665-9440665 on Build GRCh38
rs41264173 Chr.2: 9440675-9440675 on Build GRCh38
rs769888887 Chr.2: 9440572-9440572 on Build GRCh38
rs754390797 Chr.2: 9440635-9440635 on Build GRCh38
rs1004474248 Chr.2: 9440677-9440677 on Build GRCh38
rs781302767 Chr.2: 9440483-9440483 on Build GRCh38
rs573001104 Chr.2: 9440537-9440537 on Build GRCh38
rs772145566 Chr.2: 9440633-9440633 on Build GRCh38
rs151261715 Chr.2: 9440585-9440585 on Build GRCh38
rs553000464 Chr.2: 9440470-9440470 on Build GRCh38
rs760575081 Chr.2: 9440648-9440648 on Build GRCh38
rs748578851 Chr.2: 9440582-9440583 on Build GRCh38

View Full Product Details