Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56054252-56054521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284004 c.233C>G p.A78G Chr.7: 56054400-56054400 on Build GRCh38
461573 c.288C>G p.V96V Chr.7: 56054455-56054455 on Build GRCh38
3881897 c.247G>A p.A83T Chr.7: 56054414-56054414 on Build GRCh38
4801304 c.306G>A p.L102L Chr.7: 56054473-56054473 on Build GRCh38
6457739 c.244A>G p.T82A Chr.7: 56054411-56054411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777367513 Chr.7: 56054283-56054284 on Build GRCh38
rs762347825 Chr.7: 56054350-56054350 on Build GRCh38
rs1025706655 Chr.7: 56054283-56054283 on Build GRCh38
rs756442371 Chr.7: 56054310-56054310 on Build GRCh38
rs369085180 Chr.7: 56054331-56054331 on Build GRCh38
rs753333082 Chr.7: 56054476-56054476 on Build GRCh38
rs6593297 Chr.7: 56054365-56054365 on Build GRCh38
rs776703100 Chr.7: 56054429-56054429 on Build GRCh38
rs368822326 Chr.7: 56054461-56054461 on Build GRCh38
rs915028804 Chr.7: 56054410-56054410 on Build GRCh38
rs148293442 Chr.7: 56054485-56054485 on Build GRCh38
rs752436753 Chr.7: 56054370-56054370 on Build GRCh38
rs774601547 Chr.7: 56054458-56054458 on Build GRCh38
rs776592029 Chr.7: 56054349-56054349 on Build GRCh38
rs562387995 Chr.7: 56054455-56054455 on Build GRCh38
rs745638524 Chr.7: 56054405-56054405 on Build GRCh38
rs779741978 Chr.7: 56054414-56054414 on Build GRCh38
rs141399856 Chr.7: 56054491-56054491 on Build GRCh38
rs11555466 Chr.7: 56054442-56054442 on Build GRCh38
rs969752290 Chr.7: 56054356-56054356 on Build GRCh38
rs768769859 Chr.7: 56054427-56054427 on Build GRCh38
rs544183196 Chr.7: 56054443-56054443 on Build GRCh38
rs767056742 Chr.7: 56054366-56054366 on Build GRCh38
rs772266813 Chr.7: 56054318-56054318 on Build GRCh38
rs1052256300 Chr.7: 56054407-56054407 on Build GRCh38
rs200576651 Chr.7: 56054471-56054471 on Build GRCh38
rs771892723 Chr.7: 56054411-56054411 on Build GRCh38
rs747203461 Chr.7: 56054424-56054424 on Build GRCh38
rs573031496 Chr.7: 56054282-56054282 on Build GRCh38
rs750317267 Chr.7: 56054494-56054494 on Build GRCh38
rs765848090 Chr.7: 56054361-56054361 on Build GRCh38
rs755755545 Chr.7: 56054376-56054376 on Build GRCh38
rs763380767 Chr.7: 56054456-56054456 on Build GRCh38
rs935204575 Chr.7: 56054391-56054391 on Build GRCh38
rs199960641 Chr.7: 56054335-56054335 on Build GRCh38
rs377162938 Chr.7: 56054392-56054392 on Build GRCh38
rs57748693 Chr.7: 56054383-56054383 on Build GRCh38
rs748308448 Chr.7: 56054430-56054430 on Build GRCh38
rs775828022 Chr.7: 56054324-56054324 on Build GRCh38
rs778879419 Chr.7: 56054393-56054393 on Build GRCh38
rs776423297 Chr.7: 56054348-56054349 on Build GRCh38

View Full Product Details