Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67489350-67489591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIP
Entrez Gene ID
9049
Gene Name
aryl hydrocarbon receptor interacting protein
Gene Aliases
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2
UniGene
Hs.412433
Gene Chromosome Location
Chr.11: 67483029-67491108 on Build GRCh38
Transcripts
NM_001302959.1 NM_001302960.1 NM_003977.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775470 c.457G>A p.E153K Chr.11: 67489444-67489444 on Build GRCh38
4833455 c.447C>G p.I149M Chr.11: 67489434-67489434 on Build GRCh38
6045641 c.382C>T p.R128C Chr.11: 67489369-67489369 on Build GRCh38
6537424 c.450C>G p.F150L Chr.11: 67489437-67489437 on Build GRCh38
4660670 c.426G>A p.Q142Q Chr.11: 67489413-67489413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015608491 Chr.11: 67489378-67489378 on Build GRCh38
rs138312605 Chr.11: 67489402-67489402 on Build GRCh38
rs768775156 Chr.11: 67489483-67489483 on Build GRCh38
rs546909828 Chr.11: 67489560-67489560 on Build GRCh38
rs770864580 Chr.11: 67489479-67489479 on Build GRCh38
rs373159347 Chr.11: 67489464-67489464 on Build GRCh38
rs750938556 Chr.11: 67489414-67489414 on Build GRCh38
rs767830212 Chr.11: 67489413-67489413 on Build GRCh38
rs267606551 Chr.11: 67489391-67489391 on Build GRCh38
rs149570102 Chr.11: 67489404-67489404 on Build GRCh38
rs778676449 Chr.11: 67489467-67489467 on Build GRCh38
rs1004092611 Chr.11: 67489567-67489567 on Build GRCh38
rs774623231 Chr.11: 67489485-67489485 on Build GRCh38
rs750766903 Chr.11: 67489499-67489499 on Build GRCh38
rs140530307 Chr.11: 67489369-67489369 on Build GRCh38
rs199558504 Chr.11: 67489380-67489380 on Build GRCh38
rs751771656 Chr.11: 67489405-67489405 on Build GRCh38
rs1048233512 Chr.11: 67489559-67489559 on Build GRCh38
rs376193405 Chr.11: 67489444-67489444 on Build GRCh38
rs779653382 Chr.11: 67489447-67489447 on Build GRCh38
rs954223621 Chr.11: 67489415-67489415 on Build GRCh38
rs781180145 Chr.11: 67489482-67489482 on Build GRCh38
rs762219351 Chr.11: 67489406-67489406 on Build GRCh38
rs368923226 Chr.11: 67489463-67489463 on Build GRCh38
rs775549178 Chr.11: 67489393-67489393 on Build GRCh38
rs768435758 Chr.11: 67489368-67489368 on Build GRCh38
rs769856618 Chr.11: 67489391-67489391 on Build GRCh38
rs940675763 Chr.11: 67489535-67489535 on Build GRCh38
rs757087629 Chr.11: 67489546-67489546 on Build GRCh38
rs369766409 Chr.11: 67489469-67489469 on Build GRCh38
rs4084113 Chr.11: 67489566-67489566 on Build GRCh38
rs747625608 Chr.11: 67489510-67489510 on Build GRCh38
rs777413130 Chr.11: 67489529-67489529 on Build GRCh38
rs267606550 Chr.11: 67489370-67489370 on Build GRCh38
rs534510046 Chr.11: 67489545-67489545 on Build GRCh38
rs752387739 Chr.11: 67489527-67489527 on Build GRCh38
rs771591811 Chr.11: 67489544-67489544 on Build GRCh38
rs767857887 Chr.11: 67489496-67489496 on Build GRCh38
rs774280893 Chr.11: 67489512-67489512 on Build GRCh38
rs767024634 Chr.11: 67489509-67489509 on Build GRCh38
rs762009814 Chr.11: 67489486-67489486 on Build GRCh38
rs375823655 Chr.11: 67489468-67489468 on Build GRCh38
rs200995630 Chr.11: 67489475-67489475 on Build GRCh38
rs868823652 Chr.11: 67489458-67489458 on Build GRCh38
rs267606552 Chr.11: 67489411-67489411 on Build GRCh38
rs267606554 Chr.11: 67489456-67489456 on Build GRCh38
rs267606553 Chr.11: 67489416-67489416 on Build GRCh38
rs764708524 Chr.11: 67489521-67489521 on Build GRCh38
rs267607273 Chr.11: 67489471-67489471 on Build GRCh38
rs761213445 Chr.11: 67489502-67489502 on Build GRCh38
rs267607274 Chr.11: 67489470-67489470 on Build GRCh38
rs150487522 Chr.11: 67489390-67489390 on Build GRCh38

View Full Product Details