Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108046243-108046502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL5
Entrez Gene ID
8065
Gene Name
cullin 5
Gene Aliases
VACM-1, VACM1
UniGene
Hs.440320
Gene Chromosome Location
Chr.11: 108008682-108107766 on Build GRCh38
Transcripts
NM_003478.3 XM_005271682.2 XM_011543013.1 XM_017018363.1 XM_017018364.1 XM_017018365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153307 c.184C>A p.H62N Chr.11: 108046319-108046319 on Build GRCh38
7299540 c.158G>A p.W53* Chr.11: 108046293-108046293 on Build GRCh38
4675601 c.199G>T p.E67* Chr.11: 108046334-108046334 on Build GRCh38
4017575 c.188A>G p.Q63R Chr.11: 108046323-108046323 on Build GRCh38
7178549 c.231G>A p.Q77Q Chr.11: 108046366-108046366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900624446 Chr.11: 108046428-108046428 on Build GRCh38
rs145113022 Chr.11: 108046348-108046348 on Build GRCh38
rs920048047 Chr.11: 108046330-108046330 on Build GRCh38
rs77030102 Chr.11: 108046443-108046443 on Build GRCh38
rs756115807 Chr.11: 108046377-108046377 on Build GRCh38
rs938960565 Chr.11: 108046369-108046369 on Build GRCh38
rs749890524 Chr.11: 108046276-108046276 on Build GRCh38
rs947426031 Chr.11: 108046402-108046402 on Build GRCh38
rs891032377 Chr.11: 108046462-108046462 on Build GRCh38
rs374287424 Chr.11: 108046282-108046282 on Build GRCh38
rs751000380 Chr.11: 108046327-108046327 on Build GRCh38
rs757936610 Chr.11: 108046298-108046298 on Build GRCh38
rs973855568 Chr.11: 108046322-108046322 on Build GRCh38
rs141324339 Chr.11: 108046291-108046291 on Build GRCh38
rs531544413 Chr.11: 108046430-108046430 on Build GRCh38
rs948910293 Chr.11: 108046339-108046339 on Build GRCh38
rs1053024853 Chr.11: 108046378-108046378 on Build GRCh38
rs749131779 Chr.11: 108046411-108046411 on Build GRCh38
rs1008167910 Chr.11: 108046469-108046469 on Build GRCh38
rs368712400 Chr.11: 108046381-108046381 on Build GRCh38
rs7117111 Chr.11: 108046360-108046360 on Build GRCh38
rs766031826 Chr.11: 108046319-108046319 on Build GRCh38

View Full Product Details