Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118194230-118194479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAML
Entrez Gene ID
120425
Gene Name
junction adhesion molecule like
Gene Aliases
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638
UniGene
Hs.16291
Gene Chromosome Location
Chr.11: 118193727-118225094 on Build GRCh38
Transcripts
NM_001098526.1 NM_001286570.1 NM_001286571.1 NM_153206.2 NR_104479.1 XM_005271407.3 XM_017017213.1 XM_017017214.1 XM_017017215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050559 c.1066C>T p.P356S Chr.11: 118194414-118194414 on Build GRCh38
686180 c.1115A>T p.K372M Chr.11: 118194365-118194365 on Build GRCh38
5050558 c.1096C>T p.P366S Chr.11: 118194414-118194414 on Build GRCh38
5072169 c.1145A>T p.Q382L Chr.11: 118194335-118194335 on Build GRCh38
4448680 c.1138delA p.T380fs*>5 Chr.11: 118194342-118194342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019965200 Chr.11: 118194383-118194383 on Build GRCh38
rs760973212 Chr.11: 118194424-118194424 on Build GRCh38
rs774059817 Chr.11: 118194402-118194402 on Build GRCh38
rs762323830 Chr.11: 118194437-118194437 on Build GRCh38
rs757314898 Chr.11: 118194357-118194357 on Build GRCh38
rs768547342 Chr.11: 118194364-118194365 on Build GRCh38
rs376268041 Chr.11: 118194294-118194294 on Build GRCh38
rs767262200 Chr.11: 118194332-118194332 on Build GRCh38
rs776782688 Chr.11: 118194373-118194373 on Build GRCh38
rs747963271 Chr.11: 118194295-118194295 on Build GRCh38
rs373784926 Chr.11: 118194307-118194307 on Build GRCh38
rs569068918 Chr.11: 118194386-118194386 on Build GRCh38
rs188050043 Chr.11: 118194335-118194335 on Build GRCh38
rs750659401 Chr.11: 118194365-118194365 on Build GRCh38
rs369292539 Chr.11: 118194422-118194422 on Build GRCh38
rs766982029 Chr.11: 118194276-118194276 on Build GRCh38
rs764561931 Chr.11: 118194343-118194343 on Build GRCh38
rs780276395 Chr.11: 118194449-118194449 on Build GRCh38
rs780251833 Chr.11: 118194367-118194367 on Build GRCh38
rs1011217141 Chr.11: 118194382-118194382 on Build GRCh38
rs200531285 Chr.11: 118194366-118194366 on Build GRCh38
rs192635324 Chr.11: 118194438-118194438 on Build GRCh38
rs778900203 Chr.11: 118194358-118194358 on Build GRCh38
rs770575630 Chr.11: 118194395-118194395 on Build GRCh38
rs533294772 Chr.11: 118194278-118194278 on Build GRCh38
rs1002412220 Chr.11: 118194421-118194421 on Build GRCh38
rs1002661314 Chr.11: 118194411-118194411 on Build GRCh38
rs201241187 Chr.11: 118194353-118194353 on Build GRCh38
rs759108091 Chr.11: 118194412-118194412 on Build GRCh38
rs775189590 Chr.11: 118194423-118194423 on Build GRCh38
rs774149832 Chr.11: 118194313-118194313 on Build GRCh38
rs747105532 Chr.11: 118194364-118194364 on Build GRCh38
rs759203193 Chr.11: 118194322-118194322 on Build GRCh38
rs769688161 Chr.11: 118194305-118194305 on Build GRCh38
rs749180458 Chr.11: 118194394-118194394 on Build GRCh38
rs755514701 Chr.11: 118194387-118194387 on Build GRCh38
rs779131806 Chr.11: 118194274-118194274 on Build GRCh38
rs564654984 Chr.11: 118194268-118194268 on Build GRCh38
rs764481643 Chr.11: 118194434-118194434 on Build GRCh38
rs907915828 Chr.11: 118194263-118194263 on Build GRCh38
rs546676564 Chr.11: 118194312-118194312 on Build GRCh38
rs768445461 Chr.11: 118194293-118194293 on Build GRCh38
rs893410443 Chr.11: 118194373-118194376 on Build GRCh38
rs549325708 Chr.11: 118194369-118194369 on Build GRCh38

View Full Product Details