Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26638988-26639261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf71
Entrez Gene ID
285941
Gene Name
chromosome 7 open reading frame 71
Gene Aliases
-
UniGene
Hs.413394
Gene Chromosome Location
Chr.7: 26635582-26647272 on Build GRCh38
Transcripts
NM_001145531.2 NM_001302625.1 XM_017012010.1 XM_017012011.1 XM_017012012.1 XM_017012013.1 XM_017012014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762607 c.93A>G p.R31R Chr.7: 26639237-26639237 on Build GRCh38
5664279 c.21C>A p.A7A Chr.7: 26639165-26639165 on Build GRCh38
5697276 c.27C>A p.T9T Chr.7: 26639171-26639171 on Build GRCh38
5744919 c.76G>A p.A26T Chr.7: 26639220-26639220 on Build GRCh38
5026724 c.20C>G p.A7G Chr.7: 26639164-26639164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775507856 Chr.7: 26639114-26639114 on Build GRCh38
rs765756444 Chr.7: 26639165-26639165 on Build GRCh38
rs777760387 Chr.7: 26639218-26639218 on Build GRCh38
rs57000259 Chr.7: 26639237-26639237 on Build GRCh38
rs1045559838 Chr.7: 26639073-26639073 on Build GRCh38
rs901077217 Chr.7: 26639008-26639008 on Build GRCh38
rs546540623 Chr.7: 26639099-26639099 on Build GRCh38
rs535247729 Chr.7: 26639126-26639126 on Build GRCh38
rs1000101135 Chr.7: 26639094-26639094 on Build GRCh38
rs116854597 Chr.7: 26639170-26639170 on Build GRCh38
rs1022471861 Chr.7: 26639092-26639092 on Build GRCh38
rs186941328 Chr.7: 26639016-26639016 on Build GRCh38
rs902642193 Chr.7: 26639093-26639093 on Build GRCh38
rs929920470 Chr.7: 26639068-26639068 on Build GRCh38
rs866307487 Chr.7: 26639176-26639176 on Build GRCh38
rs756922035 Chr.7: 26639033-26639033 on Build GRCh38
rs566779491 Chr.7: 26639103-26639103 on Build GRCh38
rs755372116 Chr.7: 26639128-26639135 on Build GRCh38
rs937086107 Chr.7: 26639101-26639101 on Build GRCh38
rs1035230622 Chr.7: 26639114-26639114 on Build GRCh38
rs1011208840 Chr.7: 26639228-26639228 on Build GRCh38
rs1023570504 Chr.7: 26639215-26639215 on Build GRCh38
rs1049404572 Chr.7: 26639024-26639024 on Build GRCh38
rs991057431 Chr.7: 26639173-26639173 on Build GRCh38
rs754229164 Chr.7: 26639131-26639131 on Build GRCh38
rs1010631442 Chr.7: 26639050-26639050 on Build GRCh38
rs192222791 Chr.7: 26639184-26639184 on Build GRCh38
rs374358817 Chr.7: 26639185-26639185 on Build GRCh38
rs200841983 Chr.7: 26639151-26639151 on Build GRCh38

View Full Product Details