Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66566673-66566928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKLF-CMTM1
Entrez Gene ID
100529251
Gene Name
CKLF-CMTM1 readthrough
Gene Aliases
-
UniGene
Hs.698621
Gene Chromosome Location
Chr.16: 66552563-66579135 on Build GRCh38
Transcripts
NM_001202509.1 NM_001204098.1 NM_001204099.1 NM_052999.3 NM_181268.2 NM_181269.2 NM_181270.2 NM_181271.2 NM_181272.2 NM_181283.2 NM_181296.2

Gene Symbol
CMTM1
Entrez Gene ID
113540
Gene Name
CKLF like MARVEL transmembrane domain containing 1
Gene Aliases
CKLFH, CKLFH1, CKLFSF1
UniGene
Hs.15159
Gene Chromosome Location
Chr.16: 66566391-66579135 on Build GRCh38
Transcripts
NM_001202509.1 NM_001204098.1 NM_001204099.1 NM_052999.3 NM_181268.2 NM_181269.2 NM_181270.2 NM_181271.2 NM_181272.2 NM_181283.2 NM_181296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630959 c.339C>T p.R113R Chr.16: 66566852-66566852 on Build GRCh38
972177 c.389G>A p.R130H Chr.16: 66566902-66566902 on Build GRCh38
6630957 c.404C>T p.P135L Chr.16: 66566917-66566917 on Build GRCh38
1201484 c.239A>C p.K80T Chr.16: 66566752-66566752 on Build GRCh38
2993638 c.314T>C p.L105S Chr.16: 66566827-66566827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202212160 Chr.16: 66566848-66566848 on Build GRCh38
rs774480319 Chr.16: 66566867-66566867 on Build GRCh38
rs376961375 Chr.16: 66566721-66566721 on Build GRCh38
rs771383161 Chr.16: 66566851-66566851 on Build GRCh38
rs375789919 Chr.16: 66566775-66566775 on Build GRCh38
rs746594943 Chr.16: 66566908-66566908 on Build GRCh38
rs761185593 Chr.16: 66566711-66566711 on Build GRCh38
rs559055211 Chr.16: 66566827-66566828 on Build GRCh38
rs762421686 Chr.16: 66566740-66566760 on Build GRCh38
rs771748780 Chr.16: 66566839-66566839 on Build GRCh38
rs755915699 Chr.16: 66566801-66566801 on Build GRCh38
rs199641934 Chr.16: 66566770-66566770 on Build GRCh38
rs752236513 Chr.16: 66566889-66566889 on Build GRCh38
rs369937977 Chr.16: 66566763-66566763 on Build GRCh38
rs140706635 Chr.16: 66566862-66566862 on Build GRCh38
rs142981518 Chr.16: 66566909-66566909 on Build GRCh38
rs773558250 Chr.16: 66566772-66566772 on Build GRCh38
rs369671479 Chr.16: 66566796-66566796 on Build GRCh38
rs777307953 Chr.16: 66566727-66566730 on Build GRCh38
rs766799791 Chr.16: 66566713-66566713 on Build GRCh38
rs759067318 Chr.16: 66566854-66566855 on Build GRCh38
rs777158769 Chr.16: 66566854-66566854 on Build GRCh38
rs753642081 Chr.16: 66566877-66566877 on Build GRCh38
rs775637796 Chr.16: 66566863-66566863 on Build GRCh38
rs533051435 Chr.16: 66566755-66566755 on Build GRCh38
rs569712351 Chr.16: 66566714-66566714 on Build GRCh38
rs750222024 Chr.16: 66566798-66566798 on Build GRCh38
rs756076779 Chr.16: 66566732-66566732 on Build GRCh38
rs554032547 Chr.16: 66566892-66566892 on Build GRCh38
rs138390040 Chr.16: 66566878-66566878 on Build GRCh38
rs200222581 Chr.16: 66566695-66566695 on Build GRCh38
rs747614992 Chr.16: 66566837-66566837 on Build GRCh38
rs780028729 Chr.16: 66566733-66566733 on Build GRCh38
rs917016782 Chr.16: 66566749-66566749 on Build GRCh38
rs969164453 Chr.16: 66566881-66566881 on Build GRCh38
rs770430575 Chr.16: 66566861-66566861 on Build GRCh38
rs766021565 Chr.16: 66566825-66566825 on Build GRCh38
rs778333805 Chr.16: 66566819-66566819 on Build GRCh38
rs367966569 Chr.16: 66566725-66566725 on Build GRCh38
rs371441957 Chr.16: 66566739-66566739 on Build GRCh38
rs372320948 Chr.16: 66566720-66566720 on Build GRCh38
rs992537787 Chr.16: 66566742-66566742 on Build GRCh38
rs534326950 Chr.16: 66566890-66566890 on Build GRCh38
rs748865619 Chr.16: 66566690-66566690 on Build GRCh38
rs1028768131 Chr.16: 66566709-66566709 on Build GRCh38
rs376539260 Chr.16: 66566793-66566793 on Build GRCh38
rs760152037 Chr.16: 66566784-66566784 on Build GRCh38
rs907718075 Chr.16: 66566708-66566708 on Build GRCh38
rs779938973 Chr.16: 66566804-66566804 on Build GRCh38
rs754929271 Chr.16: 66566812-66566812 on Build GRCh38
rs777326414 Chr.16: 66566783-66566783 on Build GRCh38
rs753731102 Chr.16: 66566808-66566808 on Build GRCh38
rs747785103 Chr.16: 66566771-66566771 on Build GRCh38
rs376013371 Chr.16: 66566899-66566899 on Build GRCh38
rs145913875 Chr.16: 66566876-66566876 on Build GRCh38
rs761010246 Chr.16: 66566773-66566773 on Build GRCh38
rs887236642 Chr.16: 66566823-66566823 on Build GRCh38
rs368367708 Chr.16: 66566702-66566702 on Build GRCh38
rs565492845 Chr.16: 66566865-66566865 on Build GRCh38
rs145485908 Chr.16: 66566860-66566860 on Build GRCh38
rs773683378 Chr.16: 66566769-66566790 on Build GRCh38
rs76570334 Chr.16: 66566832-66566832 on Build GRCh38
rs983244190 Chr.16: 66566696-66566696 on Build GRCh38
rs765359849 Chr.16: 66566792-66566792 on Build GRCh38
rs867812910 Chr.16: 66566814-66566814 on Build GRCh38
rs768809358 Chr.16: 66566721-66566722 on Build GRCh38
rs201013566 Chr.16: 66566794-66566794 on Build GRCh38
rs975460384 Chr.16: 66566701-66566701 on Build GRCh38
rs201161463 Chr.16: 66566692-66566692 on Build GRCh38
rs770178234 Chr.16: 66566758-66566778 on Build GRCh38
rs763169929 Chr.16: 66566795-66566795 on Build GRCh38
rs763218044 Chr.16: 66566822-66566823 on Build GRCh38
rs759747563 Chr.16: 66566718-66566718 on Build GRCh38
rs201906936 Chr.16: 66566843-66566843 on Build GRCh38
rs766648884 Chr.16: 66566756-66566756 on Build GRCh38
rs765526090 Chr.16: 66566719-66566719 on Build GRCh38
rs764358984 Chr.16: 66566866-66566866 on Build GRCh38

View Full Product Details