Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129098022-129098291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAT
Entrez Gene ID
1384
Gene Name
carnitine O-acetyltransferase
Gene Aliases
CAT1
UniGene
Hs.12068
Gene Chromosome Location
Chr.9: 129094794-129110791 on Build GRCh38
Transcripts
NM_000755.3 NM_001257363.1 XM_005251708.3 XM_017014274.1 XM_017014275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3328217 c.1391G>A p.R464H Chr.9: 129098086-129098086 on Build GRCh38
1105816 c.1452C>A p.D484E Chr.9: 129098025-129098025 on Build GRCh38
5463757 c.1372C>T p.R458C Chr.9: 129098105-129098105 on Build GRCh38
3685517 c.1452C>T p.D484D Chr.9: 129098025-129098025 on Build GRCh38
1105817 c.1410G>A p.S470S Chr.9: 129098067-129098067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143682956 Chr.9: 129098104-129098104 on Build GRCh38
rs770946185 Chr.9: 129098227-129098227 on Build GRCh38
rs753149953 Chr.9: 129098160-129098160 on Build GRCh38
rs201068246 Chr.9: 129098197-129098197 on Build GRCh38
rs370320116 Chr.9: 129098245-129098245 on Build GRCh38
rs34873985 Chr.9: 129098190-129098191 on Build GRCh38
rs760054687 Chr.9: 129098153-129098153 on Build GRCh38
rs994174326 Chr.9: 129098179-129098179 on Build GRCh38
rs748031180 Chr.9: 129098204-129098204 on Build GRCh38
rs375539446 Chr.9: 129098101-129098101 on Build GRCh38
rs374018635 Chr.9: 129098185-129098185 on Build GRCh38
rs749243187 Chr.9: 129098097-129098097 on Build GRCh38
rs750039829 Chr.9: 129098266-129098266 on Build GRCh38
rs140100627 Chr.9: 129098067-129098067 on Build GRCh38
rs201613388 Chr.9: 129098188-129098188 on Build GRCh38
rs755266497 Chr.9: 129098065-129098065 on Build GRCh38
rs763445094 Chr.9: 129098248-129098248 on Build GRCh38
rs766449218 Chr.9: 129098140-129098140 on Build GRCh38
rs772385122 Chr.9: 129098128-129098128 on Build GRCh38
rs767455192 Chr.9: 129098049-129098049 on Build GRCh38
rs145973822 Chr.9: 129098191-129098191 on Build GRCh38
rs1034524171 Chr.9: 129098241-129098241 on Build GRCh38
rs774333847 Chr.9: 129098267-129098267 on Build GRCh38
rs376125366 Chr.9: 129098132-129098132 on Build GRCh38
rs200041803 Chr.9: 129098186-129098186 on Build GRCh38
rs775707790 Chr.9: 129098246-129098246 on Build GRCh38
rs982256076 Chr.9: 129098211-129098211 on Build GRCh38
rs560508389 Chr.9: 129098082-129098082 on Build GRCh38
rs773354414 Chr.9: 129098210-129098210 on Build GRCh38
rs540730402 Chr.9: 129098072-129098072 on Build GRCh38
rs16930895 Chr.9: 129098142-129098142 on Build GRCh38
rs767591349 Chr.9: 129098252-129098259 on Build GRCh38
rs376278530 Chr.9: 129098224-129098224 on Build GRCh38
rs750556155 Chr.9: 129098051-129098051 on Build GRCh38
rs368496291 Chr.9: 129098103-129098103 on Build GRCh38
rs769843161 Chr.9: 129098086-129098086 on Build GRCh38
rs372430063 Chr.9: 129098079-129098079 on Build GRCh38
rs772177459 Chr.9: 129098206-129098206 on Build GRCh38
rs769466139 Chr.9: 129098068-129098068 on Build GRCh38
rs780209200 Chr.9: 129098087-129098087 on Build GRCh38
rs776719898 Chr.9: 129098242-129098242 on Build GRCh38
rs763735366 Chr.9: 129098130-129098130 on Build GRCh38
rs202138376 Chr.9: 129098155-129098155 on Build GRCh38
rs745545439 Chr.9: 129098081-129098081 on Build GRCh38
rs761146433 Chr.9: 129098052-129098052 on Build GRCh38
rs369726339 Chr.9: 129098173-129098173 on Build GRCh38
rs367973463 Chr.9: 129098251-129098251 on Build GRCh38
rs764560591 Chr.9: 129098252-129098252 on Build GRCh38
rs146054379 Chr.9: 129098060-129098060 on Build GRCh38
rs754194294 Chr.9: 129098062-129098062 on Build GRCh38
rs756841388 Chr.9: 129098172-129098172 on Build GRCh38
rs751119057 Chr.9: 129098071-129098071 on Build GRCh38
rs754982271 Chr.9: 129098196-129098196 on Build GRCh38
rs200145368 Chr.9: 129098244-129098244 on Build GRCh38

View Full Product Details